Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: QLHYTVQEE QEHGTFVGNI AEDLGLDITK LSARGFQTVP NSRTPYLDLN LETGVLYVNE KIDREQICKQ SPSCVLHLEV FLENPLELFQ VEIEVLDIND NPPSFPEPDL TVEISESATP GTRFPLESAF DPDVGTNSLR DYEITPNSYF SLDVQTQGDG NRFAELVLEK PLDREQQAVH RYVLTAVDGG GGGGVGEGGG GGGGAGLPPQ QQRTGTALLT IRVLDSNDNV PAFDQPVYTV SLPENSPPGT LVIQLNATDP DEGQNGEVVY SFSSHISPRA RELFGLSPRT GRLEVSGELD YEESPVYQVY VQAKDLGPNA VPAHCKVLVR VLDANDNAPE ISFSTVKEAV S (19-368 aa encoded by BC111560)
This gene belongs to the protocadherin gene family, a subfamily of the cadherin superfamily. The mRNA encodes a cadherin-related neuronal receptor thought to play a role in the establishment and function of specific cell-cell connections in the brain. This family member contains 6 extracellular cadherin domains, a transmembrane domain and a cytoplasmic tail differing from those of the classical cadherins. Alternatively spliced transcripts encode isoforms with unique cytoplasmic domains.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: OL-protocadherin; ortholog of OL-pcdh; unnamed protein product
Gene Aliases: 6430521D13Rik; 6430703F07Rik; mKIAA1400; OL-pc; OL-PCDH; Olpc
UniProt ID: (Human) Q9P2E7
Entrez Gene ID: (Human) 57575, (Mouse) 18526
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support