FIGURE: 1 / 6
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
PA3-830 detects human and mouse pdx-1 alpha and shows no cross-reactivity to other pdx isoforms.
PA3-830 has been successfully used in Western blot, immunohistochemical, and immunofluorescence procedures. By Western blot, this antibody detects an ~35 kDa protein.
PA3-830 immunogen is a KLH conjugated synthetic peptide corresponding to amino acid residues 21-35 from human pdx-1 protein. This antibody is generated against the N-terminal portion of the protein, while PA3-831 is C-terminal.
The PA3-830 antibody recognizes both cytoplasmic and nuclear proteins. Formalin-fixed tissues can cause high background, so counterstaining with another antibody which is mainly expressed in the cytoplasm (i.e. insulin) is recommended.
The protein encoded by this gene is a transcriptional activator of several genes, including insulin, somatostatin, glucokinase, islet amyloid polypeptide, and glucose transporter type 2. The encoded nuclear protein is involved in the early development of the pancreas and plays a major role in glucose-dependent regulation of insulin gene expression. Defects in this gene are a cause of pancreatic agenesis, which can lead to early-onset insulin-dependent diabetes mellitus (NIDDM), as well as maturity onset diabetes of the young type 4 (MODY4).