Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • PDZRN3 Antibodies

          Abnova

          PDZRN3 Polyclonal Antibody, MaxPab™

          View all (5) PDZRN3 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite PDZRN3 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          PDZRN3 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          PDZRN3 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PDZRN3 Antibody (H00023024-B01P) in WB

          Western Blot analysis of PDZRN3 expression in transfected 293T cell line by PDZRN3 MaxPab polyclonal antibody. Lane 1: PDZRN3 transfected lysate(117.26 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PDZRN3 Antibody in Western Blot (WB)
          PDZRN3 Polyclonal Antibody, MaxPab™

          Product Details

          H00023024-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          PDZRN3 (NP_055824.1, 1 a.a. approximately 1066 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MGFELDRFDG DVDPDLKCAL CHKVLEDPLT TPCGHVFCAG CVLPWVVQEG SCPARCRGRL SAKELNHVLP LKRLILKLDI KCAYATRGCG RVVKLQQLPE HLERCDFAPA RCRHAGCGQV LLRRDVEAHM RDACDARPVG RCQEGCGLPL THGEQRAGGH CCARALRAHN GALQARLGAL HKALKKEALR AGKREKSLVA QLAAAQLELQ MTALRYQKKF TEYSARLDSL SRCVAAPPGG KGEETKSLTL VLHRDSGSLG FNIIGGRPSV DNHDGSSSEG IFVSKIVDSG PAAKEGGLQI HDRIIEVNGR DLSRATHDQA VEAFKTAKEP IVVQVLRRTP RTKMFTPPSE SQLVDTGTQT DITFEHIMAL TKMSSPSPPV LDPYLLPEEH PSAHEYYDPN DYIGDIHQEM DREELELEEV DLYRMNSQDK LGLTVCYRTD DEDDIGIYIS EIDPNSIAAK DGRIREGDRI IQINGIEVQN REEAVALLTS EENKNFSLLI ARPELQLDEG WMDDDRNDFL DDLHMDMLEE QHHQAMQFTA SVLQQKKHDE DGGTTDTATI LSNQHEKDSG VGRTDESTRN DESSEQENNG DDATASSNPL AGQRKLTCSQ DTLGSGDLPF SNESFISADC TDADYLGIPV DECERFRELL ELKCQVKSAT PYGLYYPSGP LDAGKSDPES VDKELELLNE ELRSIELECL SIVRAHKMQQ LKEQYRESWM LHNSGFRNYN TSIDVRRHEL SDITELPEKS DKDSSSAYNT GESCRSTPLT LEISPDNSLR RAAEGISCPS SEGAVGTTEA YGPASKNLLS ITEDPEVGTP TYSPSLKELD PNQPLESKER RASDGSRSPT PSQKLGSAYL PSYHHSPYKH AHIPAHAQHY QSYMQLIQQK SAVEYAQSQM SLVSMCKDLS SPTPSEPRME WKVKIRSDGT RYITKRPVRD RLLRERALKI REERSGMTTD DDAVSEMKMG RYWSKEERKQ HLVKAKEQRR RREFMMQSRL DCLKEQQAAD DRKEMNILEL SHKKMMKKRN KKIFDNWMTI QELLTHGTKS PDGTRVYNSF LSVTTV

          Target Information

          E3 ubiquitin-protein ligase. Plays an important role in regulating the surface level of MUSK on myotubes. Mediates the ubiquitination of MUSK, promoting its endocytosis and lysosomal degradation. Might contribute to terminal myogenic differentiation.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: E3 ubiquitin-protein ligase PDZRN3; Ligand of Numb protein X 3; likely ortholog of mouse semaF cytoplasmic domain associated protein 3; PDZ domain-containing RING finger protein 3; Protein SEMACAP3; RING-type E3 ubiquitin transferase PDZRN3; Semaphorin cytoplasmic domain-associated protein 3; unnamed protein product

          View more View less

          Gene Aliases: KIAA1095; LNX3; PDZRN3; SEMACAP3; SEMCAP3

          View more View less

          UniProt ID: (Human) Q9UPQ7

          View more View less

          Entrez Gene ID: (Human) 23024

          View more View less

          Function(s)
          ubiquitin-protein transferase activity zinc ion binding transferase activity metal ion binding ubiquitin protein ligase activity
          Process(es)
          neuromuscular junction development protein ubiquitination
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline