Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • PER1 Antibodies

          Abnova

          PER1 Polyclonal Antibody, MaxPab™

          View all (10) PER1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite PER1 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          PER1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          PER1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PER1 Antibody (H00005187-B01P) in WB

          Western Blot analysis of PER1 expression in transfected 293T cell line by PER1 MaxPab polyclonal antibody. Lane 1: PER1 transfected lysate(141.9 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PER1 Antibody in Western Blot (WB)

          Product Details

          H00005187-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          Immunocytochemistry (ICC/IF)

          10-100 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          PER1 (NP_002607.1, 1 a.a. approximately 1290 a.a) full-length human protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MSGPLEGADG GGDPRPGESF CPGGVPSPGP PQHRPCPGPS LADDTDANSN GSSGNESNGH ESRGASQRSS HSSSSGNGKD SALLETTESS KSTNSQSPSP PSSSIAYSLL SASSEQDNPS TSGCSSEQSA RARTQKELMT ALRELKLRLP PERRGKGRSG TLATLQYALA CVKQVQANQE YYQQWSLEEG EPCSMDMSTY TLEELEHITS EYTLQNQDTF SVAVSFLTGR IVYISEQAAV LLRCKRDVFR GTRFSELLAP QDVGVFYGST APSRLPTWGT GASAGSGLRD FTQEKSVFCR IRGGPDRDPG PRYQPFRLTP YVTKIRVSDG APAQPCCLLI AERIHSGYEA PRIPPDKRIF TTRHTPSCLF QDVDERAAPL LGYLPQDLLG APVLLFLHPE DRPLMLAIHK KILQLAGQPF DHSPIRFCAR NGEYVTMDTS WAGFVHPWSR KVAFVLGRHK VRTAPLNEDV FTPPAPSPAP SLDTDIQELS EQIHRLLLQP VHSPSPTGLC GVGAVTSPGP LHSPGSSSDS NGGDAEGPGP PAPVTFQQIC KDVHLVKHQG QQLFIESRAR PQSRPRLPAT GTFKAKALPC QSPDPELEAG SAPVQAPLAL VPEEAERKEA SSCSYQQINC LDSILRYLES CNLPSTTKRK CASSSSYTTS SASDDDRQRT GPVSVGTKKD PPSAALSGEG ATPRKEPVVG GTLSPLALAN KAESVVSVTS QCSFSSTIVH VGDKKPPESD IIMMEDLPGL APGPAPSPAP SPTVAPDPAP DAYRPVGLTK AVLSLHTQKE EQAFLSRFRD LGRLRGLDSS STAPSALGER GCHHGPAPPS RRHHCRSKAK RSRHHQNPRA EAPCYVSHPS PVPPSTPWPT PPATTPFPAV VQPYPLPVFS PRGGPQPLPP APTSVPPAAF PAPLVTPMVA LVLPNYLFPT PSSYPYGALQ TPAEGPPTPA SHSPSPSLPA LPPSPPHRPD SPLFNSRCSS PLQLNLLQLE ELPRAEGAAV AGGPGSSAGP PPPSAEAAEP EARLAEVTES SNQDALSGSS DLLELLLQED SRSGTGSAAS GSLGSGLGSG SGSGSHEGGS TSASITRSSQ SSHTSKYFGS IDSSEAEAGA ARGGAEPGDQ VIKYVLQDPI WLLMANADQR VMMTYQVPSR DMTSVLKQDR ERLRAMQKQQ PRFSEDQRRE LGAVHSWVRK GQLPRALDVM ACVDCGSSTQ DPGHPDDPLF SELDGLGLEP MEEGGGEQGS SGGGSGEGEG CEEAQGGAKA SSSQDLAMEE EEEGRSSSSP ALPTAGNCTS

          Target Information

          Circadian rhythmicity is a basic property of phylogenetically diverse organisms which range from animals and plants, to fungi. Regulation of endogenous biological clocks is regulated at the genetic level by a protein-mediated, autoregulatory feed-back loop. In mammals, several genes that encode members of the basic helix-loop helix (bHLH) PAS (PER-ARNT-SIM) transcription factor family have been shown to play a significant role in regulating circadian oscillations. Transactivation of CLOCK-induced genes is mediated via an E box enhancer (CACGTG) found upstream of target genes. CLOCK-ARNT3 heterodimers bind to E box regulatory elements and stimulate gene transcription. CLOCK has been shown to transactivate the mammalian homolog of Drosophila per. PER, in concert with the product of the mammalian timeless gene (TIM), negatively regulates its own transcription by blocking the activity of the CLOCK-BMAL1 transactivation complex.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Circadian clock protein PERIOD 1; Circadian pacemaker protein Rigui; hPer; hPER1; period circadian clock 1; Period circadian protein homolog 1; period homolog 1; Period, drosophila, homolog of; unnamed protein product

          View more View less

          Gene Aliases: hPER; KIAA0482; PER; PER1; RIGUI

          View more View less

          UniProt ID: (Human) O15534

          View more View less

          Entrez Gene ID: (Human) 5187

          View more View less

          Function(s)
          transcription regulatory region sequence-specific DNA binding RNA polymerase II core promoter proximal region sequence-specific DNA binding transcription corepressor binding protein binding kinase binding chromatin DNA binding ubiquitin protein ligase binding E-box binding DNA-binding transcription factor binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter regulation of sodium ion transport chromatin remodeling circadian rhythm entrainment of circadian clock posttranscriptional regulation of gene expression circadian regulation of gene expression regulation of hair cycle regulation of circadian rhythm negative regulation of I-kappaB kinase/NF-kappaB signaling entrainment of circadian clock by photoperiod negative regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter negative regulation of JNK cascade rhythmic process response to cAMP regulation of cytokine production involved in inflammatory response regulation of p38MAPK cascade negative regulation of glucocorticoid receptor signaling pathway
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline