Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MLEAMAEPSP EDPPPTLKPE TQPPEKRRRT IEDFNKFCSF VLAYAGYIPP SKEESDWPAS GSSSPLRGES AADSDGWDSA PSDLRTIQTF VKKAKSSKRR AAQAGPTQPG PPRSTFSRLQ APDSATLLEK MKLKDSLFDL DGPKVASPLS PTSLTHTSRP PAALTPVPLS QGDLSHPPRK KDRKNRKLGP GAGAGFGVLR RPRPTPGDGE KRSRIKKSKK RKLKKAERGD RLPPPGPPQA PPSDTDSEEE EEEEEEEEEE EMATVVGGEA PVPVLPTPPE APRPPATVHP EGVPPADSES KEVGSTETSQ DGDASSSEGE MRVMDEDIMV ESGDDSWDLI TCYCRKPFAG RPMIECSLCG TWIHLSCAKI KKTNVPDFFY CQKCKELRPE ARRLGGPPKS GEP
Acts as a negative regulator of autophagy, through promoting ubiquitination and degradation of LRSAM1, an E3 ubiquitin ligase that promotes autophagy in response to starvation or infecting bacteria.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ16355; hJUNE-1b; MGC2941; PDH-containing protein JUNE-1; PHD finger protein 23a; unnamed protein product
Gene Aliases: hJUNE-1b
UniProt ID: (Human) Q9BUL5
Entrez Gene ID: (Human) 79142
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support