Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • PLA2G6 Antibodies

          Abnova

          PLA2G6 Polyclonal Antibody, MaxPab™

          View all (14) PLA2G6 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite PLA2G6 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          PLA2G6 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          PLA2G6 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PLA2G6 Antibody (H00008398-D01P) in WB

          Western Blot analysis of PLA2G6 expression in transfected 293T cell line by PLA2G6 MaxPab polyclonal antibody. Lane 1: PLA2G6 transfected lysate(84.10 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PLA2G6 Antibody in Western Blot (WB)
          PLA2G6 Polyclonal Antibody, MaxPab™

          Product Details

          H00008398-D01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          PLA2G6 (NP_001004426.1, 1 a.a. approximately 752 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MQFFGRLVNT FSGVTNLFSN PFRVKEVAVA DYTSSDRVRE EGQLILFQNT PNRTWDCVLV NPRNSQSGFR LFQLELEADA LVNFHQYSSQ LLPFYESSPQ VLHTEVLQHL TDLIRNHPSW SVAHLAVELG IRECFHHSRI ISCANCAENE EGCTPLHLAC RKGDGEILVE LVQYCHTQMD VTDYKGETVF HYAVQGDNSQ VLQLLGRNAV AGLNQVNNQG LTPLHLACQL GKQEMVRVLL LCNARCNIMG PNGYPIHSAM KFSQKGCAEM IISMDSSQIH SKDPRYGASP LHWAKNAEMA RMLLKRGCNV NSTSSAGNTA LHVAVMRNRF DCAIVLLTHG ANADARGEHG NTPLHLAMSK DNVEMIKALI VFGAEVDTPN DFGETPTFLA SKIGRQLQDL MHISRARKPA FILGSMRDEK RTHDHLLCLD GGGVKGLIII QLLIAIEKAS GVATKDLFDW VAGTSTGGIL ALAILHSKSM AYMRGMYFRM KDEVFRGSRP YESGPLEEFL KREFGEHTKM TDVRKPKVML TGTLSDRQPA ELHLFRNYDA PETVREPRFN QNVNLRPPAQ PSDQLVWRAA RSSGAAPTYF RPNGRFLDGG LLANNPTLDA MTEIHEYNQD LIRKGQANKV KKLSIVVSLG TGRSPQVPVT CVDVFRPSNP WELAKTVFGA KELGKMVVDC CTDPDGRAVD RARAWCEMVG IQYFRLNPQL GTDIMLDEVS DTVLVNALWE TEVYIYEHRE EFQKLIQLLL SP

          Target Information

          The protein encoded by this gene is an A2 phospholipase, a class of enzyme that catalyzes the release of fatty acids from phospholipids. The encoded protein may play a role in phospholipid remodelling, arachidonic acid release, leukotriene and prostaglandin synthesis, fas-mediated apoptosis, and transmembrane ion flux in glucose-stimulated B-cells. Several transcript variants encoding multiple isoforms have been described, but the full-length nature of only two of them have been determined to date.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: 2-lysophosphatidylcholine acylhydrolase; 85 kDa calcium-independent phospholipase A2; 85/88 kDa calcium-independent phospholipase A2; CaI-PLA2; Group VI phospholipase A2; GVI PLA2; Intracellular membrane-associated calcium-independent phospholipase A2 beta; iPLA2; iPLA2-beta; neurodegeneration with brain iron accumulation 2; Palmitoyl-CoA hydrolase; Patatin-like phospholipase domain-containing protein 9; phospholipase A2, group VI (cytosolic, calcium-independent); PLA2G6; PNPLA9; unnamed protein product

          View more View less

          Gene Aliases: BB112799; CaI-PLA2; GVI; INAD1; iPLA(2)beta; iPLA2; IPLA2-VIA; iPLA2beta; NBIA2; NBIA2A; NBIA2B; PARK14; PLA2; PLA2G6; PLPLA9; PNPLA9

          View more View less

          UniProt ID: (Human) O60733, (Mouse) P97819

          View more View less

          Entrez Gene ID: (Human) 8398, (Mouse) 53357

          View more View less

          Function(s)
          1-alkyl-2-acetylglycerophosphocholine esterase activity lysophospholipase activity phospholipase A2 activity protein binding calmodulin binding hydrolase activity serine hydrolase activity identical protein binding calcium-independent phospholipase A2 activity long-chain acyl-CoA hydrolase activity protein kinase binding ATP-dependent protein binding phospholipase
          Process(es)
          lipid metabolic process chemotaxis antibacterial humoral response phosphatidylcholine catabolic process positive regulation of insulin secretion involved in cellular response to glucose stimulus cardiolipin acyl-chain remodeling Fc-gamma receptor signaling pathway involved in phagocytosis phosphatidylethanolamine catabolic process platelet activating factor metabolic process phosphatidic acid metabolic process positive regulation of ceramide biosynthetic process positive regulation of protein phosphorylation positive regulation of cytosolic calcium ion concentration memory metabolic process urinary bladder smooth muscle contraction lipid catabolic process cardiolipin biosynthetic process response to endoplasmic reticulum stress positive regulation of vasodilation positive regulation of exocytosis negative regulation of synaptic transmission, glutamatergic positive regulation of protein kinase C signaling positive regulation of release of cytochrome c from mitochondria positive regulation of arachidonic acid secretion regulation of store-operated calcium channel activity
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline