Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • PLK5 Antibodies

          Proteintech

          PLK5 Polyclonal Antibody

          3 References
          View all (7) PLK5 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite PLK5 Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (8)
          PLK5 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          PLK5 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 8

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PLK5 Antibody (21299-1-AP) in ICC/IF

          Immunofluorescent analysis of (4% PFA) fixed HeLa cells using PLK5 antibody (Product # 21299-1-AP) at dilution of 1:200 and CoraLite®488-Conjugated AffiniPure Goat Anti-Rabbit IgG(H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PLK5 Antibody in Immunocytochemistry (ICC/IF)
          PLK5 Antibody in Immunocytochemistry (ICC/IF)
          PLK5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PLK5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PLK5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PLK5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PLK5 Antibody in Western Blot (WB)
          PLK5 Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          PLK5 Polyclonal Antibody

          Product Details

          21299-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:200-1:1,000
          View 1 publication 1 publication

          Immunohistochemistry (IHC)

          -
          View 1 publication 1 publication

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:10-1:100
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          PLK5 Fusion Protein Ag15881 (1-336 aa encoded by BC100792)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.33 mg/mL

          Amount

          49.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          This antibody detects both PLK5 gene of human and mouse.

          Immunogen sequence: MYTVLTGTP PFMASPLSEM YQNIREGHYP EPAHLSANAR RLIVHLLAPN PAERPSLDHL LQDDFFTQGF TPDRLPAHSC HSPPIFAIPP PLGRIFRKVG QRLLTQCRPP CPFTPKEASG PGEGGPDPDS MEWDGESSLS AKEVPCLEGP IHLVAQGTLQ SDLAGPEGSR RPEVEAALRH LQLCLDVGPP ATQDPLGEQQ PILWAPKWVD YSSKYGFGYQ LLDGGRTGRH PHGPATPRRE GTLPTPVPPA GPGLCLLRFL ASEHALLLLF SNGMVQVSFS GVPAQLVLSG EGEGLQLTLW EQGSPGTSYS LDVPQSHGCA PTTGQHLHHA LRMLQSI (1-336 aa encoded by BC100792)

          Target Information

          Polo-like kinase 5 (PLK5) is a member of the Polo-like kinase family, notable for its involvement in cell cycle regulation processes such as mitotic entry, spindle formation, and cytoplasmic division. PLK5 is distinguished from other family members by its low expression levels, particularly within different immunophenotypes, except for being notably elevated in the C5 subtype. Its expression has been linked to poor prognosis in non-small cell lung cancer, although this is not universally supported by all data sources, suggesting complex roles in tumor biology. In a mouse model, PLK5 functions as a DNA damage-inducible gene with nucleolar localization, facilitated by a specific nucleolus localization signal in its N-terminal. The protein's ectopic expression can lead to G1 phase arrest, indicating its role in cellular response to DNA damage. Research is ongoing to fully elucidate its function and prognostic implications across various cancer types.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: polo-like kinase 5, pseudogene; serine/threonine-protein kinase PLK5; unnamed protein product

          View more View less

          Gene Aliases: 6330514A18Rik; PLK-5; PLK5P; SgK384ps

          View more View less

          UniProt ID: (Human) Q496M5, (Mouse) Q4FZD7

          View more View less

          Entrez Gene ID: (Human) 126520, (Mouse) 216166

          View more View less

          Function(s)
          protein kinase activity ATP binding kinase activity
          Process(es)
          defense response to tumor cell cellular response to DNA damage stimulus mitotic spindle organization regulation of cell cycle process positive regulation of neuron projection development regulation of apoptotic process cellular response to growth factor stimulus regulation of G1/S transition of mitotic cell cycle negative regulation of cell proliferation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-frpqk:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch