Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
This antibody is predicted to react with canine, porcine and rat based on sequence homology.
This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.
The protein encoded by this gene is a triacylglycerol lipase that mediates triacylglycerol hydrolysis in adipocytes. The encoded protein, which appears to be membrane bound, may be involved in the balance of energy usage/storage in adipocytes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: acylglycerol O-acyltransferase; acylglycerol transacylase; adiponutrin; calcium-independent phospholipase A2-epsilon; dJ549K18.1; integral membrane protein; iPLA2-epsilon; iPLA2epsilon; lysophosphatidic acid acyltransferase; patatin like phospholipase domain containing 3; patatin-like phospholipase domain-containing protein 3; unnamed protein product
Gene Aliases: ADPN; C22orf20; iPLA(2)epsilon
UniProt ID: (Human) Q9NST1, (Mouse) Q91WW7
Entrez Gene ID: (Human) 80339, (Mouse) 116939
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support