Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • PODN Antibodies

          Abnova

          PODN Polyclonal Antibody, MaxPab™

          View all (6) PODN antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite PODN Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          PODN Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          PODN Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PODN Antibody (H00127435-B01P) in WB

          Western Blot analysis of PODN expression in transfected 293T cell line by PODN MaxPab polyclonal antibody. Lane 1: PODN transfected lysate(72.71 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PODN Antibody in Western Blot (WB)

          Product Details

          H00127435-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          PODN (AAH30608, 1 a.a. approximately 661 a.a) full-length human protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MEGARARGAQ LRLGERVRPV GRRSAPGRSR FHQPWRPGAS DSAPPAGTMA QSRVLLLLLL LPPQLHLGPV LAVRAPGFGR SGGHSLSPEE NEFAEEEPVL VLSPEEPGPG PAAVSCPRDC ACSQEGVVDC GGIDLREFPG DLPEHTNHLS LQNNQLEKIY PEELSRLHRL ETLNLQNNRL TSRGLPEKAF EHLTNLNYLY LANNKLTLAP RFLPNALISV DFAANYLTKI YGLTFGQKPN LRSVYLHNNK LADAGLPDNM FNGSSNVEVL ILSSNFLRHV PKHLPPALYK LHLKNNKLEK IPPGAFSELS SLRELYLQNN YLTDEGLDNE TFWKLSSLEY LDLSSNNLSR VPAGLPRSLV LLHLEKNAIR SVDANVLTPI RSLEYLLLHS NQLREQGIHP LAFQGLKRLH TVHLYNNALE RVPSGLPRRV RTLMILHNQI TGIGREDFAT TYFLEELNLS YNRITSPQVH RDAFRKLRLL RSLDLSGNRL HTLPPGLPRN VHVLKVKRNE LAALARGALA GMAQLRELYL TSNRLRSRAL GPRAWVDLAH LQLLDIAGNQ LTEIPEGLPE SLEYLYLQNN KISAVPANAF DSTPNLKGIF LRFNKLAVGS VVDSAFRRLK HLQVLDIEGN LEFGDISKDR GRLGKEKEEE EEEEEEEEET R

          Target Information

          Non-collagenous proteins of the small leucine-rich repeat (SLR) protein family are considered important components of the extracellular matrix. The extracellular matrix plays an integral role in the pivotal processes of development, tissue repair and metastasis by regulating cell proliferation, differentiation, adhesion and migration. Podocan, also known as PCAN, SLRR5A or PODN, is a 613 amino acid secreted protein belonging to the small leucine-rich proteoglycan (SLRP) family and the class V subfamily. Expressed in kidney, heart, liver, pancreas and vascular smooth muscle cells, Podocan exists as three alternatively spliced isoforms containing twenty LRR (leucine-rich repeats), a unique N-terminal cysteine-rich cluster pattern and a highly acidic C-terminal domain. Podocan is considered a glycoprotein containing N-linked oligosaccharides that may be involved in growth regulation in cardiovascular tissues. Encoded by a gene located on human chromosome 1, Podocan interacts type I collagen (COL1).

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: class V small leucine-rich repeat protein; Podocan; podocan proteoglycan; PRO332; unnamed protein product; UNQ293

          View more View less

          Gene Aliases: PCAN; PODN; SLRR5A; UNQ293/PRO332

          View more View less

          UniProt ID: (Human) Q7Z5L7

          View more View less

          Entrez Gene ID: (Human) 127435

          View more View less

          Function(s)
          protein binding collagen binding extracellular matrix structural constituent conferring compression resistance
          Process(es)
          negative regulation of cell proliferation negative regulation of cell migration
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline