Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • PPAR alpha Antibodies

          Rockland

          PPAR alpha Polyclonal Antibody

          View all (14) PPAR alpha antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite PPAR alpha Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (8)
          PPAR alpha Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          PPAR alpha Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 8

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PPAR alpha Antibody (600-401-421) in ICC/IF

          Immunofluorescence microscopy of Rabbit Anti-PPAR alpha (N-terminal specific) antibody using (A) Mouse NIH/3T3 or (B) Human HEK293 cells fixed with MeOH. (C) Secondary antibody only with NIH/3T3 cells. Anti-PPAR alpha antibody was used at 10 µg/mL, 1h at RT. Secondary antibody: Anti-RABBIT IgG DyLight™ 488 Conjugated Preadsorbed (p/n 611-741-127) at 5 µg/mL for 1 h at RT. Staining: PPAR as green fluorescent... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PPAR alpha Antibody in Immunocytochemistry (ICC/IF)
          PPAR alpha Antibody in Immunocytochemistry (ICC/IF)
          PPAR alpha Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PPAR alpha Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PPAR alpha Antibody in Western Blot (WB)
          PPAR alpha Antibody in Western Blot (WB)
          PPAR alpha Antibody in Western Blot (WB)
          PPAR alpha Antibody in ELISA (ELISA)
          PPAR alpha Polyclonal Antibody

          Product Details

          600-401-421

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500- 1:2,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:100-1:300
          -

          Immunocytochemistry (ICC/IF)

          1-5 µg/mL
          -

          ELISA (ELISA)

          1:75,000-1:125,000
          -

          Immunoprecipitation (IP)

          1:500-1:2,000
          -
          Product Specifications

          Species Reactivity

          Bovine, Dog, Hamster, Mouse, Pig, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          PPAR alpha Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to a N-Terminal region near amino acids 1-25 of mouse PPAR alpha.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          0.02M potassium phosphate, pH 7.2, with 0.15M NaCl

          Contains

          0.01% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Store vial at -20° C prior to opening. Aliquot contents and freeze at -20° C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4° C as an undiluted liquid. Dilute only prior to immediate use.

          Anti-PPAR alpha Antibody is directed against mouse PPAR alpha protein. A BLAST analysis was used to suggest reactivity with this protein from mouse, rat, bovine, dog, golden hamster and boar sources based on 100% homology for the immunogen sequence. Cross reactivity with PPAR alpha protein from human, chimpanzee and rhesus monkey may also occur as this sequence shows 88% homology (16/18 identities) with the protein from these sources. Cross reactivity with PPAR alpha homologues from other sources has not been determined. No reactivity is expected against other subtypes of PPAR.

          Target Information

          Peroxisome proliferators are non-genotoxic carcinogens which are purported to exert their effect on cells through their interaction with members of the nuclear hormone receptor family termed peroxisome proliferator activated receptors (PPAR's). Nuclear hormone receptors are ligand-dependent intracellular proteins that stimulate transcription of specific genes by binding to specific DNA sequences following activation by the appropriate ligand. Studies indicate that PPAR's are activated by peroxisome proliferators such as clofibric acid, nafenopin, and WY-14,643, as well as by some fatty acids. It has also been shown that PPAR's can induce transcription of acyl coenzyme A oxidase & cytochrome P450 (CYP450) A6 through interaction with specific response elements. PPAR, like several other nuclear hormone receptors, heterodimerizes with retinoid X receptor (RXR) alpha.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: MGC2237; MGC2452; nuclear hormone receptor; nuclear receptor; Nuclear receptor subfamily 1 group C member 1; peroxisome proliferative activated receptor alpha; peroxisome proliferative activated receptor, alpha; peroxisome proliferator; peroxisome proliferator activated receptor; Peroxisome proliferator-activated receptor alpha; PPAR alpha; PPAR-alpha; transcription factor belonging to the nuclear receptor superfamily

          View more View less

          Gene Aliases: 4933429D07Rik; AW742785; I79_022392; NR1C1; PPAR; PPAR-alpha; PPARA; PPARALPHA

          View more View less

          UniProt ID: (Dog) Q95N78, (Rat) P37230, (Mouse) P23204

          View more View less

          Entrez Gene ID: (Pig) 397239, (Bovine) 281992, (Dog) 403654, (Rat) 25747, (Chinese hamster) 100758990, (Mouse) 19013

          View more View less

          Function(s)
          DNA binding transcription factor activity, sequence-specific DNA binding drug binding zinc ion binding lipid binding signaling receptor activity sequence-specific DNA binding RNA polymerase II core promoter proximal region sequence-specific DNA binding transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding RNA polymerase II repressing transcription factor binding transcriptional activator activity, RNA polymerase II transcription factor binding steroid hormone receptor activity receptor activity RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding protein binding phosphatase binding protein domain specific binding ubiquitin conjugating enzyme binding protein complex binding NFAT protein binding MDM2/MDM4 family protein binding metal ion binding protein heterodimerization activity C4 zinc finger nuclear receptor DNA-binding transcription factor zinc finger transcription factor gene-specific transcriptional regulator
          Process(es)
          fatty acid metabolic process epidermis development regulation of fatty acid metabolic process intracellular receptor signaling pathway negative regulation of appetite circadian regulation of gene expression regulation of circadian rhythm positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter negative regulation of transcription from RNA polymerase II promoter response to hypoxia transcription, DNA-templated regulation of transcription, DNA-templated transcription from RNA polymerase II promoter heart development regulation of gene expression negative regulation of macrophage derived foam cell differentiation negative regulation of receptor biosynthetic process negative regulation of cholesterol storage negative regulation of sequestering of triglyceride negative regulation of protein binding response to insulin behavioral response to nicotine wound healing lipoprotein metabolic process steroid hormone mediated signaling pathway positive regulation of gluconeogenesis negative regulation of blood pressure positive regulation of fatty acid oxidation enamel mineralization regulation of glycolytic process by positive regulation of transcription from RNA polymerase II promoter regulation of cellular ketone metabolic process by positive regulation of transcription from RNA polymerase II promoter regulation of lipid transport by positive regulation of transcription from RNA polymerase II promoter negative regulation of neuron death negative regulation of transcription regulatory region DNA binding glucose metabolic process lipid metabolic process rhythmic process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline