Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • PPFIBP2 Antibodies

          Abnova

          PPFIBP2 Polyclonal Antibody, MaxPab™

          View all (9) PPFIBP2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite PPFIBP2 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          PPFIBP2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          PPFIBP2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PPFIBP2 Antibody (H00008495-D01P) in WB

          Western Blot analysis of PPFIBP2 expression in transfected 293T cell line (H00008495-T02) by PPFIBP2 MaxPab polyclonal antibody.

          Lane 1: PPFIBP2 transfected lysate(98.40 KDa).
          Lane 2: Non-transfected lysate.
          {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PPFIBP2 Antibody in Western Blot (WB)
          PPFIBP2 Polyclonal Antibody, MaxPab™

          Product Details

          H00008495-D01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1-5 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          PPFIBP2 (NP_003612.1, 1 a.a. approximately 876 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          See Label

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Sequence of this protein is as follows: MASDASHALE AALEQMDGII AGTKTGADLS DGTCEPGLAS PASYMNPFPV LHLIEDLRLA LEMLELPQER AALLSQIPGP TAAYIKEWFE ESLSQVNHHS AASNETYQER LARLEGDKES LILQVSVITD QVEAQGEKIR DLEVCLEGHQ VKLNAAEEML QQELLSRTSL ETQKLDLMTE VSELKLKLVG MEKEQREQEE KQRKAEELLQ ELRHLKIKVE ELENERNQYE WKLKATKAEV AQLQEQVALK DAEIERLHSQ LSRTAALHSE SHTERDQEIQ RLKMGMETLL LANEDKDRRI EELTGLLNQY RKVKEIVMVT QGPSERTLSI NEEEPEGGFS KWNATNKDPE ELFKQEMPPR CSSPTVGPPP LPQKSLETRA QKKLSCSLED LRSESVDKCM DGNQPFPVLE PKDSPFLAEH KYPTLPGKLS GATPNGEAAK SPPTICQPDA TGSSLLRLRD TESGWDDTAV VNDLSSTSSG TESGPQSPLT PDGKRNPKGI KKFWGKIRRT QSGNFYTDTL GMAEFRRGGL RATAGPRLSR TRDSKGQKSD ANAPFAQWST ERVCAWLEDF GLAQYVIFAR QWVSSGHTLL TATPQDMEKE LGIKHPLHRK KLVLAVKAIN TKQEEKSALL DHIWVTRWLD DIGLPQYKDQ FHESRVDGRM LQYLTVNDLL FLKVTSQLHH LSIKCAIHVL HVNKFNPHCL HRRPADESNL SPSEVVQWSN HRVMEWLRSV DLAEYAPNLR GSGVHGGLII LEPRFTGDTL AMLLNIPPQK TLLRRHLTTK FNALIGPEAE QEKREKMASP AYTPLTTTAK VRPRKLGFSH FGNIRKKKFD ESTDYICPME PSDGVSDSHR VYSGYRGLSP LDAPELDGLD QVGQIS

          Target Information

          The PPFIBP2 (PPFIA Binding Protein 2) gene encodes a protein that belongs to the liprin family, which is involved in intracellular signaling and cytoskeletal organization. Located on chromosome 1, PPFIBP2 is primarily known for its role in synaptic function and cell adhesion. The protein interacts with receptor protein tyrosine phosphatases, particularly PTPRF and PTPRD, and plays a critical role in the formation and maintenance of synaptic contacts. PPFIBP2 facilitates the proper localization of synaptic vesicles and synapse-specific proteins, contributing to synaptic stability and plasticity. Disruption of PPFIBP2 expression or function has been linked to neurological disorders, emphasizing its importance in neural communication. Additionally, PPFIBP2 has been implicated in cancer, where it may influence tumor cell adhesion and migration.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: liprin beta 2; Liprin-beta-2; Protein tyrosine phosphatase receptor type f polypeptide-interacting protein-binding protein 2; PTPRF interacting protein, binding protein 2 (liprin beta 2); PTPRF-interacting protein-binding protein 2; unnamed protein product

          View more View less

          Gene Aliases: Cclp1; PPFIBP2

          View more View less

          UniProt ID: (Human) Q8ND30

          View more View less

          Entrez Gene ID: (Human) 8495

          View more View less

          Function(s)
          protein binding identical protein binding
          Process(es)
          neuromuscular junction development
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline