Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MFLTRSEYDR GVNTFSPEGR LFQVEYAIEA IKLGSTAIGI QTSEGVCLAV EKRITSPLME PSSIEKIVEI DAHIGCAMSG LIADAKTLID KARVETQNHW FTYNETMTVE SVTQAVSNLA LQFGEEDADP GAMSRPFGVA LLFGGVDEKG PQLFHMDPSG TFVQCDARAI GSASEGAQSS LQEVYHKSMT LKEAIKSSLI ILKQVMEEKL NATNIELATV QPGQNFHMFT KEELEEVIKD I
The proteasome is a multicatalytic proteinase complex with a highly ordered ring-shaped 20S core structure. The core structure is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes a member of the peptidase T1A family, that is a 20S core alpha subunit.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alpha-5; epididymis tissue sperm binding protein Li 11n; macropain subunit zeta; macropain zeta chain; multicatalytic endopeptidase complex zeta chain; proteasome (prosome, macropain) subunit, alpha type 5; proteasome (prosome, macropain) subunit, alpha type, 5; proteasome alpha 5 subunit; proteasome component 5; proteasome subunit alpha 5; proteasome subunit zeta; proteasome zeta chain; PSMA5; unnamed protein product
Gene Aliases: PSC5; ZETA
UniProt ID: (Human) P28066, (Mouse) Q9Z2U1
Entrez Gene ID: (Human) 5686, (Mouse) 26442
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support