Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • PTCH Antibodies

          Invitrogen

          PTCH Polyclonal Antibody

          View all (38) PTCH antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite PTCH Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (4)
          PTCH Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          PTCH Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 4

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PTCH Antibody (PA5-142733) in IHC (P)

          Immunohistochemistry (Paraffin) analysis of PTCH in paraffin embedded Human Cortex using PTCH Polyclonal Antibody (Product # PA5-142733) at 8 µg/mL. Heat induced antigen retrieval with citrate buffer pH 6, HRP-staining. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PTCH Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PTCH Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PTCH Antibody in Immunocytochemistry (ICC/IF)
          PTCH Antibody in Western Blot (WB)
          PTCH Polyclonal Antibody

          Product Details

          PA5-142733

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1-3 µg/mL
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          8 µg/mL
          -

          Immunocytochemistry (ICC/IF)

          5 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Goat / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Peptide with sequence C-EELKKINYQVDSWEE.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          100 µg

          Purification

          Ammonium sulfate precipitation

          Storage buffer

          TBS, pH 7.3, with 0.5% BSA

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2933379

          Product Specific Information

          Additional Information: This antibody is expected to recognize all reported isoforms (NP_000255.2; NP_001077072.1; NP_001077071.1; NP_001077073.1).

          This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.

          Target Information

          This gene encodes a member of the patched gene family. The encoded protein is the receptor for sonic hedgehog, a secreted molecule implicated in the formation of embryonic structures and in tumorigenesis, as well as the desert hedgehog and indian hedgehog proteins. This gene functions as a tumor suppressor. Mutations of this gene have been associated with basal cell nevus syndrome, esophageal squamous cell carcinoma, trichoepitheliomas, transitional cell carcinomas of the bladder, as well as holoprosencephaly. Alternative splicing results in multiple transcript variants encoding different isoforms. Additional splice variants have been described, but their full length sequences and biological validity cannot be determined currently.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: etID309849.2; FLJ26746; FLJ42602; HGNC:9585; mesenchymal dysplasia; OTTHUMP00000021709; OTTHUMP00000021710; patched; Patched 1; patched gene homolog; similar to Drosophila patched protein, Swiss-Prot Accession Number P18502; transmembrane protein; Method: conceptual translation supplied by author; patched1; Protein patched homolog 1; PTC; ptc-1; PTCH protein -3; Ptch1 protein +12b; RP11-435O5.3; unnamed protein product

          View more View less

          Gene Aliases: A230106A15Rik; BCNS; BCNS1; mes; NBCCS; PTC; PTC1; PTCH; PTCH1; SLC65B1

          View more View less

          UniProt ID: (Human) Q13635, (Mouse) Q61115

          View more View less

          Entrez Gene ID: (Human) 5727, (Mouse) 19206

          View more View less

          Function(s)
          patched binding smoothened binding protein binding hedgehog receptor activity heparin binding cholesterol binding cyclin binding macromolecular complex binding hedgehog family protein binding zinc ion binding protein complex binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter branching involved in ureteric bud morphogenesis in utero embryonic development cell fate determination neural tube formation neural tube closure heart morphogenesis signal transduction spermatid development regulation of mitotic cell cycle pattern specification process brain development negative regulation of cell proliferation epidermis development regulation of smoothened signaling pathway response to xenobiotic stimulus response to mechanical stimulus organ morphogenesis dorsal/ventral pattern formation epidermal cell fate specification response to chlorate negative regulation of keratinocyte proliferation positive regulation of cholesterol efflux protein processing spinal cord motor neuron differentiation neural tube patterning dorsal/ventral neural tube patterning neural plate axis specification embryonic limb morphogenesis prostate gland development mammary gland development response to estradiol response to retinoic acid regulation of protein localization limb morphogenesis hindlimb morphogenesis regulation of growth negative regulation of multicellular organism growth regulation of cell proliferation glucose homeostasis response to alkaloid keratinocyte proliferation positive regulation of epidermal cell differentiation negative regulation of osteoblast differentiation negative regulation of smoothened signaling pathway negative regulation of transcription, DNA-templated positive regulation of transcription, DNA-templated embryonic organ development smooth muscle tissue development epithelial cell proliferation negative regulation of epithelial cell proliferation negative regulation of cell division pharyngeal system development mammary gland duct morphogenesis mammary gland epithelial cell differentiation cell differentiation involved in kidney development somite development cellular response to cholesterol commissural neuron axon guidance stem cell proliferation cell proliferation involved in metanephros development metanephric collecting duct development protein localization to plasma membrane liver regeneration negative regulation of stem cell proliferation smoothened signaling pathway negative regulation of sequence-specific DNA binding transcription factor activity smoothened signaling pathway involved in dorsal/ventral neural tube patterning protein targeting to plasma membrane
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline