In IHC applications, antigen retrieval using Proteinase K treatment is recommended.
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Bovine, Elephant (100%) Mouse, Dog, Bat, Panda, Rabbit, Pig (94%) Rat, Hamster (89%).
PTHR2 is a specific receptor for parathyroid hormone. The activity of this receptor is mediated by g proteins which activate adenylyl cyclase. PTHR2 may be responsible for PTH effects in a number of physiological systems, and may also play a significant role in pancreatic function. PTHR2 presence in neurons indicates that it may function as a neurotransmitter receptor.