Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • RAC1/RAC2/RAC3 Antibodies

          Invitrogen

          RAC1/RAC2/RAC3 Recombinant Rabbit Monoclonal Antibody (16D14)

          View all (7) RAC1/RAC2/RAC3 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite RAC1/RAC2/RAC3 Recombinant Rabbit Monoclonal Antibody (16D14)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          RAC1/RAC2/RAC3 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          RAC1/RAC2/RAC3 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          RAC1/RAC2/RAC3 Antibody (MA572651) in WB

          Western blot analysis of RAC1/RAC2/RAC3 in Brain and HeLa lysates (25 µg/lane). Samples were incubated with RAC1/RAC2/RAC3 Recombinant Monoclonal (Product # MA572651) at 1:2,000 dilution and 4°C overnight incubation. Followed by conjugated secondary antibody incubation at room temperature for 60 min. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          RAC1/RAC2/RAC3 Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          MA572651

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Expression System

          HEK293 cells

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          16D14

          Immunogen

          KLH conjugated synthetic peptide derived from human Rac1.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.6 mg/mL

          Amount

          60 µg

          Purification

          Protein A

          Storage buffer

          TBS, pH 7.4, with 50% glycerol, 1% BSA

          Contains

          0.02% ProClin 300

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Target Information

          The protein encoded by this gene is a GTPase which belongs to the RAS superfamily of small GTP-binding proteins. Members of this superfamily appear to regulate a diverse array of cellular events, including the control of cell growth, cytoskeletal reorganization, and the activation of protein kinases. Two transcript variants encoding different isoforms have been found for this gene.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: cell migration-inducing gene 5 protein; EN-7 protein; p21-Rac1; p21-Rac2; p21-Rac3; protein EN-7; Rac1ins; RAC2; ras-like protein; ras-like protein TC25; RAS-related C3 botulinum substrate 1; RAS-related C3 botulinum substrate 2; RAS-related C3 botulinum substrate 3; ras-related C3 botulinum toxin substrate; ras-related C3 botulinum toxin substrate 1 (rho family small GTP binding protein Rac1); ras-related C3 botulinum toxin substrate 1 (rho family, small GTP binding protein Rac1); ras-related C3 botulinum toxin substrate 2 (rho family, small GTP binding protein Rac2); ras-related C3 botulinum toxin substrate 3 (rho family, small GTP binding protein Rac3); Ras-related C3 botulinum toxin substrate 3 (rho family, small GTP-binding protein Rac2); rho family, small GTP binding protein Rac1; rho family, small GTP binding protein Rac3; small G protein; small GTP binding protein; Rac1; Rho-related protein; unnamed protein product

          View more View less

          Gene Aliases: D5Ertd559e; EN-7; Gx; HSPC022; IMD73A; IMD73B; IMD73C; MIG5; MRD48; p21-Rac1; p21-Rac2; Rac-1; Rac1B; TC-25

          View more View less

          UniProt ID: (Human) P63000, (Human) P15153, (Human) P60763, (Mouse) P63001, (Rat) Q6RUV5, (Mouse) Q05144, (Mouse) P60764

          View more View less

          Entrez Gene ID: (Human) 5879, (Human) 5880, (Human) 5881, (Mouse) 19353, (Rat) 363875, (Mouse) 19354, (Rat) 366957, (Mouse) 170758, (Rat) 688319

          View more View less

          Function(s)
          GTPase activity obsolete small monomeric GTPase activity protein binding GTP binding enzyme binding protein kinase binding GTP-dependent protein binding thioesterase binding macromolecular complex binding Rho GDP-dissociation inhibitor binding protein kinase regulator activity calcium-dependent protein binding small GTPase binding histone deacetylase binding ATPase binding
          Process(es)
          chemotaxis actin filament organization establishment or maintenance of cell polarity signal transduction G-protein coupled receptor signaling pathway small GTPase mediated signal transduction positive regulation of cell proliferation regulation of cell shape positive regulation of lamellipodium assembly regulation of cell-substrate adhesion actin cytoskeleton organization cortical cytoskeleton organization regulation of actin cytoskeleton organization regulation of T cell proliferation superoxide anion generation erythrocyte enucleation regulation of mast cell degranulation bone resorption homeostasis of number of cells regulation of respiratory burst regulation of mast cell chemotaxis lymphocyte aggregation positive regulation of neutrophil chemotaxis regulation of neutrophil migration motor neuron axon guidance Wnt signaling pathway Rac protein signal transduction cell projection assembly positive regulation of cell adhesion mediated by integrin intracellular signal transduction respiratory burst cell chemotaxis postsynaptic actin cytoskeleton organization regulation of postsynapse assembly positive regulation of substrate adhesion-dependent cell spreading regulation of hydrogen peroxide metabolic process positive regulation of establishment of protein localization to mitochondrion regulation of neuron maturation cerebral cortex GABAergic interneuron development regulation of cell morphogenesis neuron projection development homeostasis of number of cells within a tissue neuromuscular process controlling balance neuromuscular process synaptic transmission, GABAergic neuron migration positive regulation of protein phosphorylation auditory receptor cell morphogenesis mast cell chemotaxis sphingosine-1-phosphate signaling pathway epithelial cell morphogenesis endocytosis phagocytosis, engulfment hyperosmotic response cytoskeleton organization cell adhesion cell-matrix adhesion enzyme linked receptor protein signaling pathway axon guidance regulation of lamellipodium assembly positive regulation of endothelial cell migration regulation of fibroblast migration negative regulation of fibroblast migration positive regulation of cell-substrate adhesion dendrite development cell migration cerebral cortex radially oriented cell migration embryonic olfactory bulb interneuron precursor migration lamellipodium assembly actin filament polymerization regulation of cell migration positive regulation of actin filament polymerization forebrain development positive regulation of microtubule polymerization response to lipopolysaccharide negative regulation of interleukin-23 production substrate adhesion-dependent cell spreading non-canonical Wnt signaling pathway positive regulation of insulin secretion involved in cellular response to glucose stimulus angiotensin-activated signaling pathway engulfment of apoptotic cell cell-cell junction organization regulation of nitric oxide biosynthetic process positive regulation of DNA replication regulation of JAK-STAT cascade hepatocyte growth factor receptor signaling pathway regulation of neuronal synaptic plasticity anatomical structure arrangement neuron projection morphogenesis dendrite morphogenesis cell motility positive regulation of filopodium assembly regulation of stress fiber assembly positive regulation of stress fiber assembly localization within membrane positive regulation of focal adhesion assembly positive regulation of protein kinase B signaling Wnt signaling pathway, planar cell polarity pathway positive regulation of dendritic spine development regulation of cell adhesion involved in heart morphogenesis regulation of ERK5 cascade cellular response to mechanical stimulus semaphorin-plexin signaling pathway dopaminergic neuron differentiation protein localization to plasma membrane cochlea morphogenesis ruffle assembly positive regulation of ruffle assembly regulation of synaptic vesicle endocytosis positive regulation of bicellular tight junction assembly midbrain dopaminergic neuron differentiation positive regulation of ovarian follicle development inflammatory response response to wounding anatomical structure morphogenesis ruffle organization positive regulation of Rho protein signal transduction negative regulation of receptor-mediated endocytosis multicellular organismal homeostasis
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-6b866bbc8c-mwps6:80/100.66.76.64:80.
          git-commit: 9d76681b284ad04e8ddcd97b79462898b32e8e5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.61.1-2026.10.99-1.0