Product References
Rad51 expression is a useful predictive factor for the efficacy of neoadjuvant chemoradiotherapy in squamous cell carcinoma of the esophagus.
Annals of surgical oncology
Nakanoko T,Saeki H,Morita M,Nakashima Y,Ando K,Oki E,Ohga T,Kakeji Y,Toh Y,Maehara Y
MA5-14416 was used in immunohistochemistry to study the value of the immunohistochemical expression of Rad51 in predicting the response of patients with squamous cell carcinoma of the esophagus to neoadjuvant chemotherapy
Sat Feb 01 00:00:00 EST 2014
Gene expression profiling reveals activation of the FA/BRCA pathway in advanced squamous cervical cancer with intrinsic resistance and therapy failure.
BMC cancer
Balacescu O,Balacescu L,Tudoran O,Todor N,Rus M,Buiga R,Susman S,Fetica B,Pop L,Maja L,Visan S,Ordeanu C,Berindan-Neagoe I,Nagy V
MA5-14416 was used in immunohistochemistry to study the gene expression profiles of patients with locally advanced squamous cervical cancer to identify pathways responsible for therapeutic failure
Tue Apr 08 00:00:00 EDT 2014
A single cohesin complex performs mitotic and meiotic functions in the protist tetrahymena.
PLoS genetics
Howard-Till RA,Lukaszewicz A,Novatchkova M,Loidl J
MA5-14416 was used in immunocytochemistry to study the ability of a single Tetrahymena cohesin complex to perform both mitotic and meiotic functions
Fri Mar 01 00:00:00 EST 2013
Nucleostemin prevents telomere damage by promoting PML-IV recruitment to SUMOylated TRF1.
The Journal of cell biology
Hsu JK,Lin T,Tsai RY
MA5-14416 was used in immunocytochemistry to study the roles of PML-IV and SUMOylated TRF1 in the mechanism by which nucleostemin protects telomeres against damage
Mon May 28 00:00:00 EDT 2012
Human papillomavirus in oral atrophic lichen planus lesions.
Oral oncology
Mattila R,Rautava J,Syrjänen S
MA5-14416 was used in immunohistochemistry to study the involvement of human papillomavirus in oral atrophic lichen planus lesions
Mon Oct 01 00:00:00 EDT 2012
The Tetrahymena meiotic chromosome bouquet is organized by centromeres and promotes interhomolog recombination.
Journal of cell science
Loidl J,Lukaszewicz A,Howard-Till RA,Koestler T
MA5-14416 was used in immunocytochemistry to study the essential role of centromere function in formation of the chromosome bouquet and inter-homolog recombination in Tetrahymena
Sat Dec 01 00:00:00 EST 2012
The recombinases Rad51 and Dmc1 play distinct roles in DNA break repair and recombination partner choice in the meiosis of Tetrahymena.
PLoS genetics
Howard-Till RA,Lukaszewicz A,Loidl J
Published figure using RAD51 monoclonal antibody (Product # MA5-14416) in Western Blot
Tue Mar 01 00:00:00 EST 2011
The role of meiotic cohesin REC8 in chromosome segregation in gamma irradiation-induced endopolyploid tumour cells.
Experimental cell research
Erenpreisa J,Cragg MS,Salmina K,Hausmann M,Scherthan H
MA5-14416 was used in immunocytochemistry to study the role of meiotic cohesin REC8 in chromosome segregation in gamma irradiation-induced endopolyploid tumour cells
Thu Sep 10 00:00:00 EDT 2009
DNA double-strand breaks, but not crossovers, are required for the reorganization of meiotic nuclei in Tetrahymena.
Journal of cell science
Mochizuki K,Novatchkova M,Loidl J
MA5-14416 was used in immunocytochemistry to study the role of DNA double-strand breaks in the reorganization of meiotic nuclei in Tetrahymena
Tue Jul 01 00:00:00 EDT 2008
Meiotic recombination proteins localize to linear elements in Schizosaccharomyces pombe.
Chromosoma
Lorenz A,Estreicher A,Kohli J,Loidl J
MA5-14416 was used in immunocytochemistry to study the localization of meiotic recombination proteins in Schizosaccharomyces pombe
Tue Aug 01 00:00:00 EDT 2006
Selenium protection from DNA damage involves a Ref1/p53/Brca1 protein complex.
Anticancer research
Fischer JL,Lancia JK,Mathur A,Smith ML
MA5-14416 was used in immunoprecipitation to study the role of the Ref1-p53-Brca1 protein complex in seleno-L-methionine-mediated protection against DNA damage
Thu May 11 00:00:00 EDT 2006
Mechanisms of enhanced radiation response following epidermal growth factor receptor signaling inhibition by erlotinib (Tarceva).
Cancer research
Chinnaiyan P,Huang S,Vallabhaneni G,Armstrong E,Varambally S,Tomlins SA,Chinnaiyan AM,Harari PM
MA5-14416 was used in western blot to study the therapeutic efficacy of epidermal growth factor receptor signaling inhibitor erlotinib and its mechanism
Fri Apr 15 00:00:00 EDT 2005
Interaction of radiation- and bleomycin-induced lesions and influence of glutathione level on the interaction.
Mutagenesis
Dutta A,Chakraborty A,Saha A,Ray S,Chatterjee A
MA5-14416 was used in western blot to study the effect of the glutathione levels on the interaction of radiation- and bleomycin-induced lesions
Thu Sep 01 00:00:00 EDT 2005
Organization and pairing of meiotic chromosomes in the ciliate Tetrahymena thermophila.
Journal of cell science
Loidl J,Scherthan H
MA5-14416 was used in immunohistochemistry to study the organization and pairing of meiotic chromosomes in the ciliate Tetrahymena thermophila
Mon Nov 15 00:00:00 EST 2004
Endopolyploid cells produced after severe genotoxic damage have the potential to repair DNA double strand breaks.
Journal of cell science
Ivanov A,Cragg MS,Erenpreisa J,Emzinsh D,Lukman H,Illidge TM
MA5-14416 was used in immunocytochemistry to study the role of endopolyploid cells in the repair of DNA double strand breaks
Wed Oct 15 00:00:00 EDT 2003
Analysis of murine Brca2 reveals conservation of protein-protein interactions but differences in nuclear localization signals.
The Journal of biological chemistry
Sarkisian CJ,Master SR,Huber LJ,Ha SI,Chodosh LA
MA5-14416 was used in western blot to compare the protein-protein interactions and nuclear localization signals of human and mouse BRCA2
Fri Oct 05 00:00:00 EDT 2001