Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • RAGE Antibodies

          Invitrogen

          RAGE Polyclonal Antibody

          View all (26) RAGE antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite RAGE Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          RAGE Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          RAGE Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          RAGE Antibody (PA5-89548) in WB

          Western blot analysis of RAGE in extracts of various cell lines. Samples were incubated with RAGE Polyclonal antibody (Product # PA5-89548) using a dilution of 1:1,000, followed by HRP Goat Anti-Rabbit IgG (H+L) at a dilution of 1:10,000. Lysates/proteins: 25 µg per lane. Blocking buffer: 3% nonfat dry milk in TBST. Detection: ECL Basic Kit. Exposure time: 90s. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          RAGE Antibody in Western Blot (WB)
          RAGE Polyclonal Antibody

          Product Details

          PA5-89548

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -

          ELISA (ELISA)

          1 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Recombinant fusion protein containing a sequence corresponding to amino acids 30-330 of human AGER (NP_0011271)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.24 mg/mL

          Amount

          24 µg

          Purification

          Affinity Chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2805643

          Product Specific Information

          Immunogen sequence: IGEPLVLKCK GAPKKPPQRL EWKLNTGRTE AWKVLSPQGG GPWDSVARVL PNGSLFLPAV GIQDEGIFRC QAMNRNGKET KSNYRVRVYQ IPGKPEIVDS ASELTAGVPN KVGTCVSEGS YPAGTLSWHL DGKPLVPNEK GVSVKEQTRR HPETGLFTLQ SELMVTPARG GDPRPTFSCS FSPGLPRHRA LRTAPIQPRV WEPVPLEEVQ LVVEPEGGAV APGGTVTLTC EVPAQPSPQI HWMKDGVPLP LPPSPVLILP EIGPQDQGTY SCVATHSSHG PQESRAVSIS IIEPGEEGPT A; Positive Samples: A-549, NCI-H460, Mouse small intestine, Rat kidney; Cellular Location: Cell membrane, Single-pass type I membrane protein

          Target Information

          The Receptor for Advanced Glycation End-products (RAGE) is a gene located on human chromosome 6p21.3, encoding a transmembrane receptor belonging to the immunoglobulin superfamily. RAGE is expressed in various tissues, with significant levels in the lungs, and plays a crucial role in cellular signaling and inflammation. As a receptor, RAGE binds multiple ligands, including advanced glycation end-products (AGEs), amyloid-beta peptide, high mobility group box 1 (HMGB1), and S100/calgranulin proteins, facilitating diverse pathological processes like inflammation, cancer progression, and neurodegeneration. The interaction between RAGE and its ligands triggers intracellular signaling pathways such as NF-kB activation, leading to inflammatory responses and oxidative stress. In the context of chronic diseases like diabetes, Alzheimer's, and cardiovascular diseases, RAGE is a critical mediator, linking metabolic disturbance to cellular dysfunction. Therapeutic targeting of RAGE signaling is under investigation, aiming to mitigate its contribution to inflammatory and degenerative diseases.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: advanced glycation end-products receptor; advanced glycosylation end product receptor; Advanced glycosylation end product-specific receptor; advanced glycosylation end product-specific receptor variant 2; advanced glycosylation end product-specific receptor variant 3; advanced glycosylation end product-specific receptor variant 4; advanced glycosylation end product-specific receptor variant 5; RAGE-4 ORF3; RAGEdelex2-6; RAGEdelex3; RAGEdelex3-10; RAGEdelex3-7; RAGEdelex8&int9; RAGEint4; RAGEint4&9; RAGEint9&delex11; RAGEv1; RAGEv8; receptor for advanced glycation end-products; receptor for advanced glycation end-products variant 20; Receptor for advanced glycosylation end products; soluble receptor; sRAGE; unnamed protein product

          View more View less

          Gene Aliases: AGER; RAGE; SCARJ1; sRAGE

          View more View less

          UniProt ID: (Human) Q15109, (Mouse) Q62151

          View more View less

          Entrez Gene ID: (Human) 177, (Rat) 81722, (Mouse) 11596

          View more View less

          Function(s)
          beta-amyloid binding DNA binding RNA binding transmembrane signaling receptor activity scavenger receptor activity laminin receptor activity protein binding signaling receptor activity histone binding identical protein binding S100 protein binding macromolecular complex binding advanced glycation end-product receptor activity binding, bridging high mobility group box 1 binding cell adhesion molecule immunoglobulin superfamily cell adhesion molecule
          Process(es)
          response to hypoxia microglial cell activation positive regulation of cytokine production regulation of T cell mediated cytotoxicity DNA replication DNA repair phagocytosis inflammatory response cellular response to DNA damage stimulus cell adhesion cell surface receptor signaling pathway learning or memory positive regulation of cell proliferation response to wounding negative regulation of biosynthetic process glucose mediated signaling pathway astrocyte development regulation of cell adhesion positive regulation of cell migration neuron projection development negative regulation of interleukin-10 production positive regulation of chemokine production positive regulation of interleukin-1 beta production positive regulation of interleukin-12 production positive regulation of interleukin-6 production positive regulation of tumor necrosis factor production positive regulation of heterotypic cell-cell adhesion positive regulation of activated T cell proliferation transcytosis positive regulation of cell differentiation positive regulation of JNK cascade astrocyte activation regulation of synaptic plasticity regulation of inflammatory response induction of positive chemotaxis positive regulation of DNA metabolic process positive regulation of NF-kappaB transcription factor activity negative regulation of multicellular organismal process positive regulation of ERK1 and ERK2 cascade cellular response to glucose stimulus positive regulation of monocyte chemotactic protein-1 production protein localization to membrane regulation of spontaneous synaptic transmission transport across blood-brain barrier regulation of long-term synaptic potentiation negative regulation of long-term synaptic potentiation negative regulation of long term synaptic depression regulation of p38MAPK cascade positive regulation of p38MAPK cascade regulation of NIK/NF-kappaB signaling positive regulation of NIK/NF-kappaB signaling positive regulation of amyloid precursor protein catabolic process negative regulation of blood circulation positive regulation of endothelin production negative regulation of connective tissue replacement involved in inflammatory response wound healing response to amyloid-beta cellular response to beta-amyloid positive regulation of DNA-dependent DNA replication positive regulation of monocyte extravasation regulation of CD4-positive, alpha-beta T cell activation positive regulation of double-strand break repair positive regulation of dendritic cell differentiation negative regulation of protein phosphorylation positive regulation of protein phosphorylation negative regulation of endothelial cell proliferation negative regulation of cell adhesion JAK-STAT cascade brain development glycoprotein metabolic process response to fructose positive regulation of autophagy negative regulation of endothelial cell migration positive regulation of gene expression positive regulation of epithelial to mesenchymal transition positive regulation of fibroblast migration response to activity positive regulation of smooth muscle cell migration lung development negative regulation of collagen biosynthetic process response to vitamin A response to genistein negative regulation of osteoblast proliferation cellular response to drug positive regulation of apoptotic process positive regulation of JUN kinase activity positive regulation of neuron apoptotic process positive regulation of fibroblast proliferation positive regulation of smooth muscle cell proliferation positive regulation of inflammatory response regulation of DNA binding response to methylglyoxal calcium ion homeostasis response to hyperoxia positive regulation of phagocytosis, engulfment transdifferentiation cellular response to hydrogen peroxide cellular response to fatty acid cellular response to organic cyclic compound response to selenite ion positive regulation of potassium ion transmembrane transporter activity positive regulation of neuron death positive regulation of endothelial cell apoptotic process positive regulation of reactive oxygen species metabolic process positive regulation of type B pancreatic cell apoptotic process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-kg5vd:80/100.66.78.64:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0