Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • RANBP6 Antibodies

          Abnova

          RANBP6 Polyclonal Antibody, MaxPab™

          View all (6) RANBP6 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite RANBP6 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          RANBP6 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          RANBP6 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          RANBP6 Antibody (H00026953-B01P) in WB

          Western Blot analysis of RANBP6 expression in transfected 293T cell line by RANBP6 MaxPab polyclonal antibody. Lane 1: RANBP6 transfected lysate(121.55 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          RANBP6 Antibody in Western Blot (WB)
          RANBP6 Polyclonal Antibody, MaxPab™

          Product Details

          H00026953-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          Immunocytochemistry (ICC/IF)

          10-100 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          RANBP6 (NP_036548, 1 a.a. approximately 1105 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5-1 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MAATASAGVP ATVSEKQEFY QLLKNLINPS CMVRRQAEEI YENIPGLCKT TFLLDAVRNR RAGYEVRQMA AALLRRLLSS GFEEVYPNLP ADVQRDVKIE LILAVKLETH ASMRKKLCDI FAVLARNLID EDGTNHWPEG LKFLIDSIYS KNVVLWEVAL HVFWHFPGIF GTQERHDLDI IKRLLDQCIQ DQEHPAIRTL SARAAAAFVL ANENNIALFK DFADLLPGIL QAVNDSCYQD DDSVLESLVE IADTVPKYLG PYLEDTLQLS LKLCGDSRLS NLQRQLALEV IVTLSETATP MLKKHTNIIA QAVPHILAMM VDLQDDEDWV NADEMEEDDF DSNAVAAESA LDRLACGLGG KVVLPMTKEH IMQMLQSPDW KYRHAGLMAL SAIGEGCHQQ MESILDETVN SVLLFLQDPH PRVRAAACTT LGQMATDFAP NFQKKFHETV IAALLRTMEN QGNQRVQSHA ASALIIFIED CPKSLLVLYV DSMVKNLHSV LVIKLQELIR NGTKLALEQL VTTIASVADT IEEKFVPYYD IFMPSLKHIV ELAVQKELKL LRGKTIECIS HIGLAVGKEK FMQDASNVMQ LLLKTQSDLN NMEDDDPQTS YMVSAWARMC KILGKDFQQY LPLVIEPLIK TASAKPDVAL LDTQDVENMS DDDGWQFVNL GDQQSFGIKT SGLEAKATAC QMLVYYAKEL REGFVEYTEQ VVKLMVPLLK FYFHDNVRVA AAESMPFLLE CARIRGPEYL AQMWQFICDP LIKAIGTEPD TDVLSEIMNS FAKSIEVMGD GCLNDEHLEE LGGILKAKLE GHFKNQELRQ VKRQEENYDQ QVEMSLQDED ECDVYILTKV SDILHSLFST YKEKILPWFE QLLPLIVNLI CSSRPWPDRQ WGLCIFDDII EHCSPTSFKY VEYFRWPMLL NMRDNNPEVR QAAAYGLGVM AQFGGDDYRS LCSEAVPLLV KVIKCANSKT KKNVIATENC ISAIGKILKF KPNCVNVDEV LPHWLSWLPL HEDKEEAIQT LSFLCDLIES NHPVVIGPNN SNLPKIISII AEGKINETIN YEDPCAKRLA NVVRQVQTSE DLWLECVSQL DDEQQEALQE LLNFA

          Target Information

          RANBP6 (Ran-binding protein 6) belongs to the importin beta family and may function in nuclear protein import as a nuclear transport receptor.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Ran BP6; Ran-binding protein 6; Ran-BP6; Ran-GTP binding protein; RanBP6; unnamed protein product

          View more View less

          Gene Aliases: RANBP6

          View more View less

          UniProt ID: (Human) O60518

          View more View less

          Entrez Gene ID: (Human) 26953

          View more View less

          Function(s)
          protein binding nuclear localization sequence binding nuclear import signal receptor activity
          Process(es)
          protein import into nucleus protein transport
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-msbq7:80/100.66.78.64:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0