Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • RANBP9 Antibodies

          Proteintech

          RANBP9 Polyclonal Antibody

          4 References
          View all (14) RANBP9 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite RANBP9 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (30)
          RANBP9 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          RANBP9 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 30

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          RANBP9 Antibody (17755-1-AP) in ICC/IF

          Immunofluorescent analysis of HeLa cells using 17755-1-AP (RANBP9 antibody) at dilution of 1:50 and Alexa Fluor 488-conjugated AffiniPure Goat Anti-Rabbit IgG (H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          RANBP9 Antibody in Immunocytochemistry (ICC/IF)
          RANBP9 Antibody in Immunocytochemistry (ICC/IF)
          RANBP9 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RANBP9 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RANBP9 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RANBP9 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RANBP9 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RANBP9 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RANBP9 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RANBP9 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RANBP9 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RANBP9 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RANBP9 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RANBP9 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RANBP9 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RANBP9 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RANBP9 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RANBP9 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RANBP9 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RANBP9 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RANBP9 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RANBP9 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RANBP9 Antibody in Western Blot (WB)
          RANBP9 Antibody in Western Blot (WB)
          RANBP9 Antibody in Western Blot (WB)
          RANBP9 Antibody in Western Blot (WB)
          RANBP9 Antibody in Western Blot (WB)
          RANBP9 Antibody in Western Blot (WB)
          RANBP9 Antibody in Western Blot (WB)
          RANBP9 Antibody in Immunoprecipitation (IP)
          RANBP9 Polyclonal Antibody

          Product Details

          17755-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          View 2 publications 2 publications

          Immunohistochemistry (IHC)

          -
          View 1 publication 1 publication

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:20-1:200
          View 1 publication 1 publication

          Immunoprecipitation (IP)

          0.5-4.0 µg
          -
          Product Specifications

          Species Reactivity

          Dog, Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          RANBP9 Fusion Protein Ag12112 (1-389 aa encoded by BC063849)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.21 mg/mL

          Amount

          31.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: MREWRTKIQ AQIDRFPIGD REGEWQTMIQ KMVSSYLVHH GYCATAEAFA RSTDQTVLEE LASIKNRQRI QKLVLAGRMG EAIETTQQLY PSLLERNPNL LFTLKVRQFI EMVNGTDSEV RCLGGRSPKS QDSYPVSPRP FSSPSMSPSH GMNIHNLASG KGSTAHFSGF ESCSNGVISN KAHQSYCHSN KHQSSNLNVP ELNSINMSRS QQVNNFTSND VDMETDHYSN GVGETSSNGF LNGSSKHDHE MEDCDTEMEV DSSQLRRQLC GGSQAAIERM IHFGRELQAM SEQLRRDCGK NTANKKMLKD AFSLLAYSDP WNSPVGNQLD PIQREPVCSA LNSAILETHN LPKQPPLALA MGQATQCLGL MARSGIGSCA FATVEDYLH (1-389 aa encoded by BC063849 )

          Target Information

          This gene encodes a protein that binds RAN, a small GTP binding protein belonging to the RAS superfamily that is essential for the translocation of RNA and proteins through the nuclear pore complex. The protein encoded by this gene has also been shown to interact with several other proteins, including met proto-oncogene, homeodomain interacting protein kinase 2, androgen receptor, and cyclin-dependent kinase 11.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: B cell antigen receptor Ig beta associated protein 1; B-cell antigen receptor Ig beta-associated protein 1; BPM 90; BPM L; BPM-L; BPM90; BPML; IBAP 1; IBAP-1; Imp 9; Importin 9; novel centrosomal protein RanBPM; RAN binding protein 9; Ran Binding Protein in the Microtubule organizing center; ran binding protein, centrosomal; Ran BP9; Ran-binding protein 9; Ran-binding protein M; RanBP 7; RANBP 9; RanBP7; RanBP9; RanBPM; unnamed protein product

          View more View less

          Gene Aliases: BPM-L; BPM90; IBAP-1; Ibap1; RanBP7; RANBP9; RANBPM; RGD1561940

          View more View less

          UniProt ID: (Human) Q96S59, (Mouse) P69566

          View more View less

          Entrez Gene ID: (Human) 10048, (Dog) 478728, (Mouse) 56705, (Rat) 364686

          View more View less

          Function(s)
          protein binding enzyme binding small GTPase binding Ran GTPase binding scaffold/adaptor protein
          Process(es)
          cytoskeleton organization microtubule nucleation protein-containing complex assembly negative regulation of ERK1 and ERK2 cascade positive regulation of amyloid precursor protein catabolic process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-msbq7:80/100.66.78.64:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0