Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: GKAEELHYP LGERRSDYDR EALLGVQEDV DEYVKLGHEE QQKRLQAIIK KIDLDSDGFL TESELSSWIQ MSFKHYAMQE AKQQFVEYDK NSDDTVTWDE YNIQMYDRVI DFDENTALDD AEEESFRKLH LKDKKRFEKA NQDSGPGLSL EEFIAFEHPE EVDYMTEFVI QEALEEHDKN GDGFVSLE (23-209 aa encoded by BC004892 )
The protein encoded by this gene is a calcium-binding protein located in the lumen of the ER. The protein contains six conserved regions with similarity to a high affinity Ca(+2)-binding motif, the EF-hand. This gene maps to the same region as type 4 Bardet-Biedl syndrome, suggesting a possible causative role for this gene in the disorder. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: calcium-binding protein ERC-55; E6-binding protein; reticulocalbin 2, EF-hand calcium binding domain; reticulocalbin 2, EF-hand calcium binding domain (endoplasmic reticulum calcium-binding protein, 55kD); taipoxin-associated calcium binding protein 49; taipoxin-associated calcium-binding protein 49; TCBP-49; TCBP49; unnamed protein product
Gene Aliases: E6BP; ERC-55; ERC55; TCBP49
UniProt ID: (Human) Q14257, (Mouse) Q8BP92, (Rat) Q62703
Entrez Gene ID: (Human) 5955, (Mouse) 26611, (Rat) 29218
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support