Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MGQQISDQT QLVINKLPEK VAKHVTLVRE SGSLTYEEFL GRVAELNDVT AKVASGQEKH LLFEVQPGSD SSAFWKVVVR VVCTKINKSS GIVEASRIMN LYQFIQLYKD ITSQAAGVLA QSSTSEEPDE NSSSVTSCQA SLWMGRVKQL TDEEECCICM DGRADLILPC AHSFCQKCID KWSDRHRNCP ICRLQMTGAN ESWVVSDAPT EDDMANYILN MADEAGQPHR P (1-230 aa encoded by BC018104)
The protein encoded by this gene contains a RING finger, a motif known to be involved in protein-DNA and protein-protein interactions. Abundant expression of this gene was found in the testicular tissue of fertile men, but was not detected in azoospermic patients. Studies of the mouse counterpart suggest that this gene may function as a testis specific transcription factor during spermatogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C3HC4-like zinc finger protein; RFP141; Ring Finger Protein 141; unnamed protein product; ZFP26; zinc finger protein 230; ZNF230
Gene Aliases: 2610110L04Rik; RFP141; ZFP26; ZFP36; ZNF230
UniProt ID: (Human) Q8WVD5, (Mouse) Q99MB7
Entrez Gene ID: (Human) 50862, (Mouse) 67150
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support