Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • ROC1 Antibodies

          Invitrogen

          ROC1 Recombinant Rabbit Monoclonal Antibody (R2D3)

          View all (13) ROC1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite ROC1 Recombinant Rabbit Monoclonal Antibody (R2D3)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (15)
          ROC1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          ROC1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 15

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          ROC1 Antibody (MA572231) in ICC/IF

          Immunocytochemistry analysis of ROC1 in HeLa cells. Permeabilized with Trinton X-100 at room temp for 20 min. Samples were incubated with ROC1 Recombinant Monoclonal (Product # MA572231 using a dilution at 1:100, for 90 min at 37°C. Followed by conjugated Goat Anti-Rabbit IgG antibody (green) at 37°C for 90 min, DAPI was used to stain the cell nuclei. PBS instead of the primary antibody was used as the blank control. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          ROC1 Antibody in Immunocytochemistry (ICC/IF)
          ROC1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ROC1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ROC1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ROC1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ROC1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ROC1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ROC1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ROC1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ROC1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ROC1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ROC1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ROC1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ROC1 Antibody in Western Blot (WB)
          ROC1 Antibody in Flow Cytometry (Flow)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          MA572231

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:100-1:500
          -

          Immunohistochemistry (Frozen) (IHC (F))

          1:100-1:500
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:200
          -

          Flow Cytometry (Flow)

          1:50-1:100
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Expression System

          HEK293 cells

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          R2D3

          Immunogen

          KLH conjugated synthetic peptide derived from human RBX1.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.26 mg/mL

          Amount

          26 µg

          Purification

          Protein A

          Storage buffer

          TBS, pH 7.4, with 50% glycerol, 1% BSA

          Contains

          0.02% ProClin 300

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Target Information

          This locus encodes a RING finger-like domain-containing protein. The encoded protein interacts with cullin proteins and likely plays a role in ubiquitination processes necessary for cell cycle progression. This protein may also affect protein turnover. Related pseudogenes exist on chromosomes 2 and 5.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: E3 ubiquitin-protein transferase RBX1; RBX1; regulator of cullins 1; RING finger protein 75; ring-box 1, E3 ubiquitin protein ligase; RING-box protein 1; unnamed protein product; ZYP protein

          View more View less

          Gene Aliases: 1500002P15Rik; BA554C12.1; RNF75; ROC1

          View more View less

          UniProt ID: (Human) P62877, (Mouse) P62878

          View more View less

          Entrez Gene ID: (Human) 9978, (Mouse) 56438, (Rat) 300084

          View more View less

          Function(s)
          ubiquitin-protein transferase activity protein binding zinc ion binding NEDD8 transferase activity ubiquitin protein ligase binding ubiquitin-ubiquitin ligase activity binding, bridging RNA polymerase II sequence-specific DNA binding transcription factor binding ubiquitin protein ligase activity NEDD8 ligase activity cullin family protein binding eukaryotic initiation factor 4E binding macromolecular complex binding
          Process(es)
          G1/S transition of mitotic cell cycle negative regulation of transcription from RNA polymerase II promoter MAPK cascade protein polyubiquitination transcription-coupled nucleotide-excision repair nucleotide-excision repair regulation of transcription, DNA-templated ubiquitin-dependent protein catabolic process protein monoubiquitination cellular iron ion homeostasis cellular response to DNA damage stimulus spermatogenesis regulation of mitotic cell cycle cell proliferation regulation of autophagy regulation of cell cycle process regulation of centrosome duplication neural crest cell differentiation protein processing protein ubiquitination regulation of DNA-dependent DNA replication initiation regulation of BMP signaling pathway SCF-dependent proteasomal ubiquitin-dependent protein catabolic process replication fork processing regulation of TOR signaling positive regulation of proteasomal ubiquitin-dependent protein catabolic process negative regulation of type I interferon production regulation of natural killer cell activation cellular response to oxidative stress cellular response to UV regulation of gene expression, epigenetic T cell activation regulation of cell proliferation regulation of circadian rhythm signal transduction in response to DNA damage regulation of apoptotic process positive regulation of I-kappaB kinase/NF-kappaB signaling negative regulation of I-kappaB kinase/NF-kappaB signaling proteasome-mediated ubiquitin-dependent protein catabolic process post-translational protein modification protein K27-linked ubiquitination protein neddylation positive regulation of cilium assembly positive regulation of protein catabolic process negative regulation of smoothened signaling pathway regulation of embryonic development negative regulation of insulin receptor signaling pathway regulation of inflammatory response centrosome duplication regulation of cell cycle limb development cilium morphogenesis regulation of gene silencing by miRNA cellular response to chemical stress protein K63-linked ubiquitination protein K48-linked ubiquitination cellular response to amino acid stimulus regulation of cellular response to stress negative regulation of canonical Wnt signaling pathway ubiquitin-dependent protein catabolic process via the C-end degron rule pathway RNA polymerase II transcription initiation surveillance negative regulation of beige fat cell differentiation regulation of cellular response to insulin stimulus regulation of mitophagy negative regulation of macromitophagy regulation of cell cycle phase transition regulation of mitotic cytokinesis positive regulation of protein autoubiquitination negative regulation of response to oxidative stress negative regulation of adipose tissue development positive regulation of TORC1 signaling regulation of xenophagy ribosome-associated ubiquitin-dependent protein catabolic process regulation of DNA damage checkpoint regulation of stem cell population maintenance negative regulation of DNA-dependent DNA replication autophagosome assembly mitophagy epithelial to mesenchymal transition transcription from RNA polymerase II promoter transcription elongation from RNA polymerase II promoter apoptotic process response to oxidative stress lysosome organization insulin receptor signaling pathway negative regulation of autophagy positive regulation of autophagy protein catabolic process DNA damage response, signal transduction by p53 class mediator cellular response to nutrient levels negative regulation of proteasomal ubiquitin-dependent protein catabolic process positive regulation of type I interferon production cellular response to insulin stimulus cellular response to amino acid starvation mRNA cleavage involved in gene silencing by miRNA p38MAPK cascade TORC1 signaling positive regulation of translation positive regulation of transcription from RNA polymerase II promoter type I interferon signaling pathway base-excision repair, AP site formation via deaminated base removal negative regulation of signal transduction by p53 class mediator positive regulation of epithelial cell apoptotic process negative regulation of TORC1 signaling negative regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-7fb9f846c9-xq5gc:80/100.66.76.64:80.
          git-commit: 8207b5afa5c305bd40e0a4f0b9b53fd46f2a8829
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.59.1-2026.09.91-1.0