    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Proteins & Peptides ›
          • SLMAP Proteins
          Invitrogen
          Human SLMAP Control Fragment Recombinant Protein
          View all (2) SLMAP proteins
          Tech Support
          Cite Human SLMAP Control Fragment Recombinant Protein
          Additional Information:
          {{banner.description}}
          Product Details
          RP-102134
          Applications
          Tested Dilution
          Publications
          Neutralization (Neu)
          Assay-dependent
          -
          Product Specifications
          Species
          Human
          Expression System
          E. coli
          Amino acid sequence
          TTDAQMDEQDLNEPLAKVSLLKALLEEERKAYRNQVEESTKQIQVLQAQLQRLHIDTENLREEKDSEITSTRDELL
          Tag
          His-ABP-tag
          Class
          Recombinant
          Type
          Protein
          Purity
          >80% by SDS-PAGE and Coomassie blue staining
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          ≥5.0 mg/mL
          Purification
          purified
          Storage buffer
          1M urea/PBS, pH 7.4
          Contains
          no preservative
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Wet ice
          Product Specific Information
          Highest antigen sequence indentity to the following orthologs: Mouse (88%), Rat (88%).
          This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-51707. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
          Target Information
          Sarcolemmal membrane associated protein (SLMAP) is a tail-anchored membrane protein involved in the docking and membrane fusion process via protein-protein interactions. Deregulation of SLMAP expression plays an important role in type 2 diabetes, and changes in SLMAP expression are linked to hyperglycemia and diabetes. SLMAPs are components of cardiac membranes and have been identified as a novel regulator of cardiac function. There are several isoforms of SLMAPs with varying molecular size (35, 45, 63, and 83-91 kDa) as a result of alternative splicing. The short 35- kDa isoform is predominantly expressed in cardiac and slow-twitch skeletal muscle, while the 83- to 91- kDa isoform is ubiquitous.
          Bioinformatics
          Protein Aliases: Sarcolemmal membrane-associated protein; Sarcolemmal-associated protein; SLAP1-Alt; unnamed protein product
          View more View less
          Gene Aliases: KIAA1601; SLAP; SLMAP; UNQ1847/PRO3577
          View more View less
          UniProt ID: (Human) Q14BN4
          View more View less
          Entrez Gene ID: (Human) 7871
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline