Search
Search
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MEGQSVEELL AKAEQDEAEK LQRITVHKEL ELQFDLGNLL ASDRNPPTGL RCAGPTPEAE LQALARDNTQ LLINQLWQLP TERVEEAIVA RLPEPTTRLP REKPLPRPRP LTRWQQFARL KGIRPKKKTN LVWDEVSGQW RRRWGYQRAR DDTKEWLIEV PGNADPLEDQ FAKRIQAKKE RVAKNELNRL RNLARAHKMQ LPSAAGLHPT GHQSKEELGR AMQVAKVSTA SVGRFQERLP KEKVPRGSGK KRKFQPLFGD FAAEKKNQLE LLRVMNSKKP QLDVTRATNK QMREEDQEEA AKRRKMSQKG KRKGGRQGPG GKRKGGPPSQ GGKRKGGLGG KMNSGPPGLG GKRKGGQRPG GKRRK
Involved in ribosomal large subunit assembly. May regulate the localization of the 5S RNP/5S ribonucleoprotein particle to the nucleolus. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: homolog of yeast ribosome biogenesis regulator; homolog of yeast ribosome biogenesis regulatory protein RRS1; KIAA0112; ribosome biogenesis regulator homolog; Ribosome biogenesis regulatory protein homolog; ribosome biogenesis regulatory protein RRS1 homolog; RRS1 ribosome biogenesis regulator homolog
Gene Aliases: KIAA0112; RRR; RRS1
UniProt ID: (Human) Q15050
Entrez Gene ID: (Human) 23212
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support