Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Renin Antibodies

          Proteintech

          Renin Polyclonal Antibody

          22 References
          View all (41) Renin antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Renin Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (14)
          Renin Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Renin Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 14

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Renin Antibody (14291-1-AP) in ICC/IF

          Immunofluorescent analysis of HepG2 cells, using REN antibody 14291-1-AP at 1:50 dilution and Rhodamine-labeled goat anti-rabbit IGG (red). Blue pseudocolor = DAPI (fluorescent DNA dye). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Renin Antibody in Immunocytochemistry (ICC/IF)
          Renin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Renin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Renin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Renin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Renin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Renin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Renin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Renin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Renin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Renin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Renin Antibody in Western Blot (WB)
          Renin Antibody in Western Blot (WB)
          Renin Antibody in Western Blot (WB)
          Renin Polyclonal Antibody

          Product Details

          14291-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          View 8 publications 8 publications

          Immunohistochemistry (IHC)

          -
          View 11 publications 11 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:400-1:1,600
          -

          Immunocytochemistry (ICC/IF)

          1:20-1:200
          View 3 publications 3 publications
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Renin Fusion Protein Ag5612 (1-406 aa encoded by BC047752)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.17 mg/mL

          Amount

          25.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: MDGWRRMPR WGLLLLLWGS CTFGLPTDTT TFKRIFLKRM PSIRESLKER GVDMARLGPE WSQPMKRLTL GNTTSSVILT NYMDTQYYGE IGIGTPPQTF KVVFDTGSSN VWVPSSKCSR LYTACVYHKL FDASDSSSYK HNGTELTLRY STGTVSGFLS QDIITVGGIT VTQMFGEVTE MPALPFMLAE FDGVVGMGFI EQAIGRVTPI FDNIISQGVL KEDVFSFYYN RDSENSQSLG GQIVLGGSDP QHYEGNFHYI NLIKTGVWQI QMKGVSVGSS TLLCEDGCLA LVDTGASYIS GSTSSIEKLM EALGAKKRLF DYVVKCNEGP TLPDISFHLG GKEYTLTSAD YVFQESYSSK KLCTLAIHAM DIPPPTGPTW ALGATFIRKF YTEFDRRNNR IGFALAR (1-406 aa encoded by BC047752)

          Target Information

          Renin is a highly specific endopeptidase, whose only known function is to generate angiotensin I from angiotensinogen in the plasma, initiating a cascade of reactions that produce an elevation of blood pressure and increased sodium retention by the kidney.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: angiotensin-forming enzyme; Angiotensinogenase; aspartyl-protease; EC 3.4.23; EC 3.4.23.15; FLJ10761; Kidney renin; REN; Renin; renin 1 structural; renin b; renin precursor, renal; Renin-1; unnamed protein product

          View more View less

          Gene Aliases: ADTKD4; D19352; HNFJ2; RATRENAA; REN; Ren-1; Ren-A; Ren1; Ren1c; Ren1d; RENAA; Rn-1; Rnr; RTD

          View more View less

          UniProt ID: (Human) P00797, (Rat) P08424, (Mouse) P06281

          View more View less

          Entrez Gene ID: (Human) 5972, (Rat) 24715, (Mouse) 19701

          View more View less

          Function(s)
          endopeptidase activity aspartic-type endopeptidase activity receptor binding insulin-like growth factor receptor binding protein binding peptidase activity hydrolase activity
          Process(es)
          kidney development angiotensin maturation regulation of blood volume by renin-angiotensin renin-angiotensin regulation of aldosterone production proteolysis establishment of blood-nerve barrier regulation of blood pressure hormone-mediated signaling pathway response to organic substance response to drug drinking behavior regulation of MAPK cascade cell maturation response to cAMP response to cGMP mesonephros development male gonad development response to lipopolysaccharide cellular response to drug response to immobilization stress beta-amyloid metabolic process regulation of blood vessel size by renin-angiotensin response to xenobiotic stimulus gene expression angiotensin-activated signaling pathway cellular response to xenobiotic stimulus juxtaglomerular apparatus development
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline