Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • S100A11 Antibodies

          Proteintech

          S100A11 Polyclonal Antibody

          59 References
          View all (24) S100A11 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite S100A11 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (23)
          S100A11 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          S100A11 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 23

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          S100A11 Antibody (10237-1-AP) in ICC/IF

          Immunofluorescent analysis of (-20°C Ethanol) fixed PC-3 cells using S100A11 antibody (Product # 10237-1-AP) at dilution of 1:400 and CoraLite®488-Conjugated AffiniPure Goat Anti-Rabbit IgG(H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          S100A11 Antibody in Immunocytochemistry (ICC/IF)
          S100A11 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          S100A11 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          S100A11 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          S100A11 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          S100A11 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          S100A11 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          S100A11 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          S100A11 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          S100A11 Antibody in Western Blot (WB)
          S100A11 Antibody in Western Blot (WB)
          S100A11 Antibody in Western Blot (WB)
          S100A11 Antibody in Western Blot (WB)
          S100A11 Antibody in Western Blot (WB)
          S100A11 Antibody in Western Blot (WB)
          S100A11 Antibody in Western Blot (WB)
          S100A11 Antibody in Western Blot (WB)
          S100A11 Antibody in Western Blot (WB)
          S100A11 Antibody in Western Blot (WB)
          S100A11 Antibody in Western Blot (WB)
          S100A11 Antibody in Western Blot (WB)
          S100A11 Antibody in Flow Cytometry (Flow)
          S100A11 Antibody in Immunoprecipitation (IP)
          S100A11 Polyclonal Antibody

          Product Details

          10237-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000-1:4,000
          View 30 publications 30 publications

          Immunohistochemistry (IHC)

          -
          View 13 publications 13 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:500
          -

          Immunocytochemistry (ICC/IF)

          1:400
          View 12 publications 12 publications

          Flow Cytometry (Flow)

          0.2 µg/1x10^6 cells
          -

          Immunoprecipitation (IP)

          0.5-4.0 µg
          View 2 publications 2 publications

          Miscellaneous PubMed (Misc)

          -
          View 2 publications 2 publications
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          S100A11 Fusion Protein Ag0343 (3-105 aa encoded by BC001410)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.46 mg/mL

          Amount

          69 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          This S100A11 antibody (10237-1-AP) has also been instrumental in investigations into S100A11's role in the development of brain tumors in tuberous sclerosis complex (TSC) patients.

          Immunogen sequence: KISSPTETE RCIESLIAVF QKYAGKDGYN YTLSKTEFLS FMNTELAAFT KNQKDPGVLD RMMKKLDTNS DGQLDFSEFL NLIGGLAMAC HDSFLKAVPS QKRT (3-105 aa encoded by BC001410)

          Target Information

          The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein may function in motility, invasion, and tubulin polymerization. Chromosomal rearrangements and altered expression of this gene have been implicated in tumor metastasis.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Calgizzarin; calizzarin; EMAP; endothelial monocyte activating polypeptide 1; Endothelial monocyte-activating polypeptide; epididymis secretory protein Li 43; Metastatic lymph node gene 70 protein; MLN 70; MLN 70, S100 C; Protein S100-A11; Protein S100-C; Protein S100C; S100 calcium binding protein A11 (calizzarin); S100 calcium binding protein A14; S100 calcium-binding protein A11; unnamed protein product

          View more View less

          Gene Aliases: cal; Emap1; EMAPI; HEL-S-43; MLN70; S100A11; S100a14; S100C

          View more View less

          UniProt ID: (Human) P31949, (Rat) Q6B345, (Mouse) P50543

          View more View less

          Entrez Gene ID: (Human) 6282, (Rat) 445415, (Mouse) 20195

          View more View less

          Function(s)
          calcium ion binding protein binding protein homodimerization activity S100 protein binding metal ion binding calcium-dependent protein binding cadherin binding involved in cell-cell adhesion calmodulin-related
          Process(es)
          signal transduction negative regulation of DNA replication negative regulation of cell proliferation positive regulation of smooth muscle cell migration regulation of cell proliferation cell-cell adhesion spermatogenesis
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-6479c7f679-26whj:80/100.66.74.142:80.
          git-commit: b70c0dceb888abc3b0dcebb5bc3a5b8753e7e816
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.50.0-Offline