    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Secondary Antibodies ›
          • Goat anti-Rabbit IgG Secondary Antibodies
          Invitrogen
          F(ab)-Goat anti-Rabbit IgG (H+L) Superclonal™ Secondary Antibody
          Advanced Verification
          View all (247) Goat anti-Rabbit IgG antibodies
          Questions & Answers
          Cite F(ab)-Goat anti-Rabbit IgG (H+L) Superclonal™ Secondary Antibody
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (4)
          • Advanced Verification (1)
          Rabbit IgG (H+L) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Rabbit IgG (H+L) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 5
          Rabbit IgG (H+L) Secondary Antibody (SA5-10395) in ICC/IF
          Immunofluorescence analysis of F(ab)-Goat anti-Rabbit IgG (H+L) Superclonal™ Recombinant Secondary Antibody (Product # SA5-10395) was performed on fixed and permeabilized HeLa cells blocked with SuperBlock™ Blocking Buffer at room temperature and stained with beta Tubulin Chimeric Recombinant Rabbit Monoclonal Antibody (S11B) (Product # MA5-47948, 1:200) in SuperBlock™ Blocking Buffer (Product # 37515, 1:20... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Rabbit IgG (H+L) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Secondary Antibody in Western Blot (WB)
          Rabbit IgG (H+L) Secondary Antibody in ELISA (ELISA)
          Rabbit IgG (H+L) Secondary Antibody in ELISA (ELISA)
          Rabbit IgG (H+L) Secondary Antibody
          Product Details
          SA5-10395
          Applications
          Tested Dilution
          Western Blot (WB)
          1:1,000
          Immunocytochemistry (ICC/IF)
          1:4,000-1:16,000
          ELISA (ELISA)
          1:1,000
          Product Specifications
          Species Reactivity
          Rabbit
          Host/Isotype
          Goat / IgG, F(ab)
          Expression System
          Expi293
          Class
          Recombinant Superclonal
          Type
          Secondary Antibody
          Immunogen
          Purified Rabbit IgG
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          250 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.4
          Contains
          0.09% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Wet ice
          Target
          IgG
          Antibody Form
          F(ab) Fragment
          Product Specific Information
          F(ab)-Goat anti-Rabbit IgG (H+L) Superclonal™ Recombinant Secondary Antibody can detect both Rabbit Fab2 and Fc fragment of Rabbit IgG. It showed no cross-reactivity to various species immunoglobulins tested.
          This Fab fragment antibody is expressed in vitro and purified as a recombinant construct. It is also engineered to deliver enhanced stability and sensitivity. The advantages of recombinant antibodies include-better specificity, animal origin-free formulation, and lot-to-lot consistency.
          F(ab) fragments offer several advantages over full-length IgGs due to their smaller molecular size (~50 kDa vs. ~150 kDa), such as improved tissue penetration. The absence of the Fc region reduces nonspecific binding and background staining in Fc receptor-expressing cells and tissues, while also enabling increased flexibility in multiplexing assays.
          It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
          Target Information
          Anti-Rabbit secondary antibodies are affinity-purified antibodies with well-characterized specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
          View product citations for antibody SA5-10395 on CiteAb
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline