FIGURE: 1 / 2
Store as concentrated solution.
Centrifuge briefly prior to opening vial.
The nonstructural protein 8 (nsp8) is one of the SARS-Coronavirus replicase cleaving products, encoded by ORF1a. Nsp8 is thought to be part of the viral replication complex, which is associated with intracellular membranes. No specific information on the function of nsp8 is available.
Protein Aliases: CoV-2; non-structural protein 8; NSP8