Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • SARS-CoV-2 Spike Protein RBD Antibodies

          Invitrogen

          SARS-CoV-2 Spike Protein (RBD) Chimeric Recombinant Rabbit Monoclonal Antibody (T01KHuRb)

          Advanced Verification
          3 References
          View all (181) SARS-CoV-2 Spike Protein RBD antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite SARS-CoV-2 Spike Protein (RBD) Chimeric Recombinant Rabbit Monoclonal Antibody (T01KHuRb)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (9)
          • Advanced Verification (2)
          SARS-CoV-2 Spike Protein (RBD) Chimeric Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          SARS-CoV-2 Spike Protein (RBD) Chimeric Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 11

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          SARS-CoV-2 Spike Protein (RBD) Chimeric Antibody (703959) in ICC/IF

          For Immunofluorescence analysis, HEK-293 cells were transfected with a vector encoding full length spike protein from SARS CoV2. Cells were fixed and permeabilized for detection of spike protein using Anti-SARS CoV2 Spike Protein RBD Recombinant Rabbit Chimera Monoclonal Antibody (Product # 703959) at a concentration of 5 µg/mL and labeled with Goat anti-Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Anti... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          SARS-CoV-2 Spike Protein (RBD) Chimeric Antibody in Immunocytochemistry (ICC/IF)
          SARS-CoV-2 Spike Protein (RBD) Chimeric Antibody in Flow Cytometry (Flow)
          SARS-CoV-2 Spike Protein (RBD) Chimeric Antibody in ELISA (ELISA)
          SARS-CoV-2 Spike Protein (RBD) Chimeric Antibody in ELISA (ELISA)
          SARS-CoV-2 Spike Protein (RBD) Chimeric Antibody in ELISA (ELISA)
          SARS-CoV-2 Spike Protein (RBD) Chimeric Antibody in ELISA (ELISA)
          SARS-CoV-2 Spike Protein (RBD) Chimeric Antibody in ELISA (ELISA)
          SARS-CoV-2 Spike Protein (RBD) Chimeric Antibody in Neutralization (Neu)
          SARS-CoV-2 Spike Protein (RBD) Chimeric Antibody
          SARS-CoV-2 Spike Protein (RBD) Chimeric Antibody
          SARS-CoV-2 Spike Protein (RBD) Chimeric Antibody

          Product Details

          703959

          Applications
          Tested Dilution
          Publications

          Immunocytochemistry (ICC/IF)

          5 µg/mL
          View 2 publications 2 publications

          Flow Cytometry (Flow)

          0.06 µg/test
          -

          ELISA (ELISA)

          0.2-2,000 ng/mL
          View 1 publication 1 publication

          Neutralization (Neu)

          0.1 - 10 µg/mL
          -
          Product Specifications

          Species Reactivity

          Virus

          Published species

          Human, Virus

          Host/Isotype

          Rabbit / IgG, kappa

          Expression System

          Expi293

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          T01KHuRb

          Immunogen

          This antibody was developed using computationally derived variants of SARS-CoV antibody structures and is shown to bind Spike Protein S1 subunit (RBD) of SARS CoV2

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.2-7.4

          Contains

          0.09% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Wet ice

          RRID

          AB_2866478

          Product Specific Information

          This antibody has a human Fab containing a kappa light chain, and Rabbit Fc. It recognizes the RBD domain of SARS CoV2 Spike protein.

          It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          This antibody is not recommended for the Western blot application.

          Target Information

          SARS-CoV-2 (Severe Acute Respiratory Syndrome Coronavirus 2) also known as 2019-nCoV (2019 Novel Coronavirus) is a virus that causes illnesses ranging from the common cold symptoms to severe effects including death and respiratory distress. The CoV Spike (S) protein plays the most important roles in viral attachment, fusion and entry, and serves as a target for development of antibodies, entry inhibitors and vaccines. Of note is the Spike protein receptor-binding domain (RBD, S-RBD) in SARS-CoV-2 Spike protein which binds strongly to human and bat angiotensin-converting enzyme 2 (ACE2) receptors. SARS-CoV-2 RBD exhibits significantly higher binding affinity to ACE2 receptor than SARS-CoV RBD in comparison. S1 contains a receptor-binding domain (RBD) that can specifically bind to angiotensin-converting enzyme 2 (ACE2), the receptor on target cells.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Coronavirus; COVID; Covid 19; COVID-19; COVID19; SARS; SARS Coronavirus; SARS CoV; SARS CoV-2; SARS CoV2; SARS Spike Protein; SARS-CoV-2; SARS-CoV2

          View more View less

          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-794c4b9cd6-97vht:80/100.66.77.199:80.
          git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.52.0-Offline