Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • SETMAR Antibodies

          Abnova

          SETMAR Polyclonal Antibody, MaxPab™

          View all (7) SETMAR antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite SETMAR Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          SETMAR Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          SETMAR Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          SETMAR Antibody (H00006419-B01P) in WB

          Western Blot analysis of SETMAR expression in transfected 293T cell line by SETMAR MaxPab polyclonal antibody. Lane 1: SETMAR transfected lysate(38.72 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          SETMAR Antibody in Western Blot (WB)

          Product Details

          H00006419-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          SETMAR (AAH11635.1, 1 a.a. approximately 352 a.a) full-length human protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MAEFKEKPEA PTEQLDVACG QENLPVGAWP PGAAPAPFQY TPDHVVGPGA DIDPTQITFP GCICVKTPCL PGTCSCLRHG ENYDDNSCLR DIGSGGKYAE PVFECNVLCR CSDHCRNRVV QKGLQFHFQV FKTHKKGWGL RTLEFIPKGR FVCEYAGEVL GFSEVQRRIH LQTKSDSNYI IAIREHVYNG QVMETFVDPT YIGNIGRFLN HSCEPNLLMI PVRIDSMVPK LALFAAKDIV PEEELSYDYS GRYLNLTVSE DKERLDHGKL RKPCYCGAKS CTAFLPFDSS LYCPVEKSNI SCGNEKEPSM CGSAPSVFPS CKRLTLEVSL FSDKQLAPPY SGRQWLASFT SA

          Target Information

          SETMAR (SET domain and mariner transposase fusion gene) is a histone methyltransferase that methylates Lys-4 and Lys-36 of histone H3, creating specific tags for epigenetic transcriptional activation. It specifically mediates dimethylation of H3 Lys-36. SETMAR has sequence-specific DNA-binding activity, and recognizes the 19-mer core of the 5'-terminal inverted repeats (TIRs) of the Hsmar1 transposable element. SETMAT has DNA nicking activity, and in vivo end-joining activity, and may mediate genomic integration of foreign DNA.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Histone-lysine N-methyltransferase SETMAR; methylase and endonuclease; Metnase; orf; encodes putative chimeric protein with SET domain in N-terminus with similarity to several other human, Drosophila, nematode and yeast proteins; SET domain and mariner transposase fusion gene-containing protein; SET domain and mariner transposase fusion protein; unnamed protein product

          View more View less

          Gene Aliases: Mar1; METNASE; SETMAR

          View more View less

          UniProt ID: (Human) Q53H47

          View more View less

          Entrez Gene ID: (Human) 6419

          View more View less

          Function(s)
          single-stranded DNA endodeoxyribonuclease activity nucleic acid binding DNA binding double-stranded DNA binding single-stranded DNA binding catalytic activity nuclease activity endonuclease activity protein binding methyltransferase activity zinc ion binding protein-lysine N-methyltransferase activity transferase activity hydrolase activity histone methyltransferase activity histone methyltransferase activity (H3-K4 specific) protein homodimerization activity DNA topoisomerase binding metal ion binding histone methyltransferase activity (H3-K36 specific) histone H3 methyltransferase activity histone H3K36 dimethyltransferase activity
          Process(es)
          DNA double-strand break processing DNA repair double-strand break repair via nonhomologous end joining DNA catabolic process chromatin organization chromatin remodeling cellular response to DNA damage stimulus cell proliferation DNA integration replication fork processing methylation mitotic DNA integrity checkpoint nucleic acid metabolic process positive regulation of DNA topoisomerase (ATP-hydrolyzing) activity positive regulation of double-strand break repair via nonhomologous end joining negative regulation of chromosome organization
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline