Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • SIRT2 Antibodies

          Fabgennix

          SIRT2 Polyclonal Antibody

          View all (35) SIRT2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite SIRT2 Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          SIRT2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          SIRT2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          SIRT2 Antibody (SIR2-201AP) in WB

          WB of SIR2-201AP with PC-SIR2. 1:500 antibody dilution in DiluObuffer. Apparent MW is 41 kDa. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          SIRT2 Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          SIRT2 Polyclonal Antibody

          Product Details

          SIR2-201AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          ELISA (ELISA)

          1:10,000
          -

          Immunoprecipitation (IP)

          1:50-1:250
          -
          Product Specifications

          Species Reactivity

          Bovine, Chicken, Human, Mouse, Non-human primate, Pig, Rat, Xenopus, Zebrafish

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic peptide corresponding to amino acids 270-287 of Rat SIR2/SIRT2 protein.Sequence:MDFDSKKAYRDVAWLGDC
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5-1.5 mg/mL

          Purification

          Affinity chromatography

          Storage buffer

          proprietary buffer, pH 7.4-7.8, with 30% glycerol, 0.5% BSA

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Target Information

          The SIRT2 protein (also known as Sirtuin for Silent Mating Type Information 2-Homolog) is a NAD-dependent deacytylase (NDAC) that has been shown to control gene silencing, cell cycle, and DNA damage repair. It is believed that SIRT2 may act as a tumor suppressor in human gliomas and may also serve as a novel molecular marker for these cells. SIRT2 has also been shown to act as a redox sensor to help regulate muscle gene expression in response to food intake and exercise. SIRT2 acts in the phosphorylation cascade involving mitosis where SIRT2 is phosphorylated in late G2 phase, during M phase, and into cytokinesis.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: 5E5 antigen; mSIR2L2; NAD-dependent deacetylase SIRT2; NAD-dependent deacetylase sirtuin-2; NAD-dependent protein deacetyalse sirtuin-2; NAD-dependent protein deacylase sirtuin-2; NAD-dependent protein defatty-acylase sirtuin-2; regulatory protein SIR2 homolog 2; silent information regulator 2; silent mating type information regulation 2, (S.cerevisiae, homolog)-like; sirtuin 2; SIR2-h; SIR2-like protein 2; sir2-related protein type 2; sir2-related protein type 2; sirtuin type 2; NAD-dependent deacetylase sirtuin-2; silent information regulator 2; SIR2-like protein 2; regulatory protein SIR2; Sirt2; silent mating type information regulation 2; transcript variant 3; NAD-dependent deacetylase sirtuin-2; mSIR2L2; SIR2-like protein 2; NAD-dependent protein deacetylase sirtuin-2; sirtuin type 2; sirtuin-like protein 2; unnamed protein product; yeast silent information regulator 2 (Sir2p) homolog

          View more View less

          Gene Aliases: 5730427M03Rik; SIR2; SIR2L; SIR2L2; wu:fb57d05; zgc:55966; zgc:77003

          View more View less

          UniProt ID: (Human) Q8IXJ6, (Mouse) Q8VDQ8, (Zebrafish) Q7ZVK3

          View more View less

          Entrez Gene ID: (Bovine) 504463, (Human) 22933, (Pig) 100125964, (Chicken) 548628, (Mouse) 64383, (Rat) 361532, (Zebrafish) 322309

          View more View less

          Function(s)
          chromatin binding NAD+ ADP-ribosyltransferase activity histone deacetylase activity protein binding zinc ion binding NAD-dependent histone deacetylase activity protein deacetylase activity NAD-dependent protein deacetylase activity histone acetyltransferase binding histone deacetylase binding tubulin deacetylase activity ubiquitin binding NAD-dependent histone deacetylase activity (H4-K16 specific) NAD binding NAD+ binding histone methacryllysine demethacrylase activity histone benzoyllysine debenzoylase activity DNA-binding transcription factor binding NAD-dependent protein demyristoylase activity NAD-dependent protein depalmitoylase activity NAD-dependent protein lysine deacylase activity protein ADP-ribosylase activity transcription factor binding transferase activity metal ion binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter chromatin silencing at rDNA chromatin organization protein deacetylation ubiquitin-dependent protein catabolic process fatty acid biosynthetic process negative regulation of autophagy negative regulation of peptidyl-threonine phosphorylation lipid catabolic process myelination in peripheral nervous system regulation of myelination positive regulation of proteasomal ubiquitin-dependent protein catabolic process cellular response to oxidative stress peptidyl-lysine deacetylation regulation of gene expression, epigenetic negative regulation of protein catabolic process regulation of apoptotic process negative regulation of apoptotic process proteasome-mediated ubiquitin-dependent protein catabolic process positive regulation of DNA binding post-translational protein modification regulation of fat cell differentiation negative regulation of fat cell differentiation negative regulation of fatty acid biosynthetic process positive regulation of fatty acid biosynthetic process positive regulation of meiotic nuclear division negative regulation of striated muscle tissue development negative regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter negative regulation of oligodendrocyte differentiation regulation of cell cycle positive regulation of cell division positive regulation of attachment of spindle microtubules to kinetochore cellular response to caloric restriction negative regulation of oligodendrocyte progenitor proliferation cellular response to hypoxia cellular response to epinephrine stimulus tubulin deacetylation regulation of postsynaptic neurotransmitter receptor internalization positive regulation of execution phase of apoptosis positive regulation of oocyte maturation negative regulation of NLRP3 inflammasome complex assembly negative regulation of reactive oxygen species metabolic process response to cocaine regulation of cell differentiation negative regulation of developmental process chromatin remodeling mitotic nuclear envelope reassembly regulation of exit from mitosis substantia nigra development heterochromatin assembly telomeric heterochromatin assembly regulation of phosphorylation regulation of lipid biosynthetic process regulation of multicellular organismal process response to redox state negative regulation of satellite cell differentiation negative regulation of cell proliferation negative regulation of angiogenesis cilium morphogenesis negative regulation of transcription from RNA polymerase II promoter in response to hypoxia positive regulation of proteasomal ubiquitin-dependent protein catabolic process involved in cellular response to hypoxia
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b6cf8ccb7-5t5fd:80/100.66.72.150:80.
          git-commit: 76b2fce788d0185f5e22c122d85c407206860d42
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.0-2026-offline-prod