Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • GLT-1 Antibodies

          Osenses

          SLC1A2 Polyclonal Antibody

          1 Reference
          View all (32) GLT-1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite SLC1A2 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}

          Product Details

          OSE00005G-500UG

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          10-50 µg/mL
          -

          Immunohistochemistry (IHC)

          10-50 µg/mL
          -

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthetic peptide from mouse SLC1A2 conjugated to blue carrier protein was used as the antigen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Concentration

          Conc. Not Determined

          Amount

          500 µg

          Purification

          Ammonium sulfate precipitation

          Storage buffer

          PBS

          Contains

          no preservative

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Glycerol (1:1) may be added for added stability.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Reconstitute in 500 µL of sterile water. Centrifuge to remove any insoluble material.

          Specificity of this antibody: EAAT2.

          Target Information

          Excitatory amino acid transporter 2 (EAAT2) is a member of a family of solute transporter proteins. The membrane-bound protein is the principal transporter that clears the excitatory neurotransmitter glutamate from the extracellular space at synapses in the central nervous system. Glutamate clearance is necessary for proper synaptic activation and to prevent neuronal damage from excessive activation of glutamate receptors. Mutations in and decreased expression of this protein are associated with amyotrophic lateral sclerosis (ALS.) Alternatively spliced transcript variants of this gene have been identified.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: EAAT2; EAAT2 variant mEAAT2/5UT5; Excitatory amino acid transporter 2; excitotoxic amino acid transporter 2; glial high affinity glutamate transporter; GLT-1; GLT1a; GLT1b; GLT1c; GLUT-R; glutamate transporter 1c; glutamate transporter-1; Glutamate/aspartate transporter II; high affinity glucose transporter; human brain glutamate transporter; OTTHUMP00000165209; OTTHUMP00000165210; Sodium-dependent glutamate/aspartate transporter 2; solute carrier family 1 (glial high affinity glutamate transporter), member 2; Solute carrier family 1 member 2; solute carrier family 1, member 2; unnamed protein product

          View more View less

          Gene Aliases: 1700091C19Rik; 2900019G14Rik; AI159670; DEE41; EAAT2; EIEE41; Glt; GLT-1; GLT1; HBGT; MGLT1; SLC1A2

          View more View less

          UniProt ID: (Human) P43004, (Mouse) P43006, (Rat) P31596

          View more View less

          Entrez Gene ID: (Human) 6506, (Mouse) 20511, (Rat) 29482

          View more View less

          Function(s)
          L-glutamate transmembrane transporter activity high-affinity glutamate transmembrane transporter activity protein binding anion transmembrane transporter activity neutral amino acid transmembrane transporter activity symporter activity glutamate:sodium symporter activity cysteine transmembrane transporter activity metal ion binding sodium:dicarboxylate symporter activity primary active transporter
          Process(es)
          glutathione biosynthetic process ion transport neurotransmitter transport amino acid transport chemical synaptic transmission nervous system development visual behavior response to xenobiotic stimulus response to light stimulus response to wounding neutral amino acid transport L-glutamate transport telencephalon development adult behavior multicellular organism growth response to amino acid positive regulation of glucose import protein homotrimerization transepithelial transport L-aspartate transmembrane transport D-aspartate import cellular response to cocaine anion transmembrane transport import into cell L-glutamate import across plasma membrane neurotransmitter reuptake L-aspartate import across plasma membrane transport across blood-brain barrier cysteine transmembrane transport transport dicarboxylic acid transport multicellular organism aging cellular response to extracellular stimulus sodium ion transmembrane transport response to drug L-glutamate import L-glutamate transmembrane transport
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline