Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • SMC4 Antibodies

          Abnova

          SMC4L1 Polyclonal Antibody, MaxPab™

          View all (15) SMC4 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite SMC4L1 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          SMC4L1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          SMC4L1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          SMC4L1 Antibody (H00010051-B01P) in WB

          Western Blot analysis of SMC4 expression in transfected 293T cell line by SMC4 MaxPab polyclonal antibody. Lane 1: SMC4L1 transfected lysate(141.68 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          SMC4L1 Antibody in Western Blot (WB)
          SMC4L1 Polyclonal Antibody, MaxPab™

          Product Details

          H00010051-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          SMC4L1 (AAI48797.1, 1 a.a. approximately 1288 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MPRKGTQPST ARRREEGPPP PSPDGASSDA EPEPPSGRTE SPATAAETAS EELDNRSLEE ILNSIPPPPP PAMTNEAGAP RLMITHIVNQ NFKSYAGEKI LGPFHKRFSC IIGPNGSGKS NVIDSMLFVF GYRAQKIRSK KLSVLIHNSD EHKDIQSCTV EVHFQKIIDK EGDDYEVIPN SNFYVSRTAC RDNTSVYHIS GKKKTFKDVG NLLRSHGIDL DHNRFLILQG EVEQIAMMKP KGQTEHDEGM LEYLEDIIGC GRLNEPIKVL CRRVEILNEH RGEKLNRVKM VEKEKDALEG EKNIAIEFLT LENEIFRKKN HVCQYYIYEL QKRIAEMETQ KEKIHEDTKE INEKSNILSN EMKAKNKDVK DTEKKLNKIT KFIEENKEKF TQLDLEDVQV REKLKHATSK AKKLEKQLQK DKEKVEEFKS IPAKSNNIIN ETTTRNNALE KEKEKEEKKL KEVMDSLKQE TQGLQKEKES REKELMGFSK SVNEARSKMD VAQSELDIYL SRHNTAVSQL TKAKEALIAA SETLKERKAA IRDIEGKLPQ TEQELKEKEK ELQKLTQEET NFKSLVHDLF QKVEEAKSSL AMNRSRGKVL DAIIQEKKSG RIPGIYGRLG DLGAIDEKYD VAISSCCHAL DYIVVDSIDI AQECVNFLKR QNIGVATFIG LDKMAVWAKK MTEIQTPENT PRLFDLVKVK DEKIRQAFYF ALRDTLVADN LDQATRVAYQ KDRRWRVVTL QGQIIEQSGT MTGGGSKVMK GRMGSSLVIE ISEEEVNKME SQLQNDSKKA MQIQEQKVQL EERVVKLRHS EREMRNTLEK FTASIQRLIE QEEYLNVQVK ELEANVLATA PDKKKQKLLE ENVSAFKTEY DAVAEKAGKV EAEVKRLHNT IVEINNHKLK AQQDKLDKIN KQLDECASAI TKAQVAIKTA DRNLQKAQDS VLRTEKEIKD TEKEVDDLTA ELKSLEDKAA EVVKNTNAAE ESLPEIQKEH RNLLQELKVI QENEHALQKD ALSIKLKLEQ IDGHIAEHNS KIKYWHKEIS KISLHPIEDN PIEEISVLSP EDLEAIKNPD SITNQIALLE ARCHEMKPNL GAIAEYKKKE ELYLQRVAEL DKITYERDSF RQAYEDLRKQ RLNEFMAGFY IITNKLKENY QMLTLGGDAE LELVDSLDPF SEGIMFSVRP PKKSWKKIFN LSGGEKTLSS LALVFALHHY KPTPLYFMDE IDAALDFKNV SIVAFYIYEQ TKNAQFIIIS LRNNMFEISD RLIGIYKTYN ITKSVAVNPK EIASKGLC

          Target Information

          The proteins belonging to structural maintenance of chromosomes (SMC) family play critical roles in chromosome structural changes. SMC proteins are involved in the proper condensation and segregation of mitotic chromosomes. Four human SMC proteins that form two distinct heterodimeric complexes in the cell, the human chromosome-associated protein (hCAP)-C and hCAP-E protein complex (hCAP-C/hCAP-E), and the human SMC1 (hSMC1) and hSMC3 protein complex (hSMC1/hSMC3) have been identified.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Chromosome-associated polypeptide C; hCAP-C; SMC protein 4; SMC4 structural maintenance of chromosomes 4-like 1; structural maintenance of chromosomes (SMC) family member; Structural maintenance of chromosomes protein 4; unnamed protein product; XCAP-C homolog

          View more View less

          Gene Aliases: CAP-C; CAPC; SMC-4; SMC4; SMC4L1

          View more View less

          UniProt ID: (Human) Q9NTJ3

          View more View less

          Entrez Gene ID: (Human) 10051

          View more View less

          Function(s)
          nucleotide binding chromatin binding single-stranded DNA binding protein binding ATP binding ATPase activity
          Process(es)
          mitotic sister chromatid segregation nuclear division mitotic chromosome condensation meiotic chromosome condensation chromosome condensation meiotic chromosome segregation chromosome organization cell division kinetochore organization positive regulation of chromosome segregation nuclear chromosome segregation positive regulation of chromosome separation positive regulation of chromosome condensation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-b97mm:80/100.66.72.101:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0