Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MPRSSRSPG DPGALLEDVA HNPRPRRIAQ RGRNTSRMAE DTSPNMNDNI LLPVRNNDQA LGLTQCMLGC VSWFTCFACS LRTQAQQVLF NTCRCKLLCQ KLMEKTGILL LCAFGFWMFS IHLPSKMKVW QDDSINGPLQ SLRLYQEKVR HHSGEIQDLR GSMNQLIAKL QEMEAMSDEQ KMAQKIMKMI HGDYIEKPDF ALKSIGASID FEHTSVTYNH EKAHSYWNWI QLWNYAQPPD VILEPNVTPG NCWAFEGDRG QVTIQLAQKV YLSNLTLQHI PKTISLSGSL DTAPKDFVIY GMEGSPKEEV FLGAFQFQPE NIIQMFPLQN QPARAFSAVK VKISSNWGNP GFTCLYRVRV HGSVAPPREQ PHQNPYPKRD (1-379 aa encoded by BC026118)
The function of this protein remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: epididymis secretory sperm binding protein; nuclear membrane protein; sad1 and UNC84 domain-containing protein 5; sperm-associated antigen 4-like protein; spermatogenesis-related gene 4; SRG4; testis and spermatogenesis related gene 4; testis and spermatogenesis-related gene 4 protein; TSARG4; unnamed protein product
Gene Aliases: 1700021O15Rik; dJ726C3.1; SPAG4L; SPAG4L-2; SPGF16; SRG4; TSARG4
UniProt ID: (Human) Q8TC36, (Mouse) Q9DA32
Entrez Gene ID: (Human) 140732, (Mouse) 76407
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support