Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MRAWIFFLL CLAGRALAAP QQEALPDETE VVEETVAEVT EVSVGANPVQ VEVGEFDDGA EETEEEVVAE NPCQNHHCKH GKVCELDENN TPMCVCQDPT SCPAPIGEFE KVCSNDNKTF DSSCHFFATK CTLEGTKKGH KLHLDYIGPC KYIPPCLDSE LTEFPLRMRD WLKNVLVTLY ERDEDNNLLT EKQKLRVKKI HENEKRLEAG DHPVELLARD FEKNYNMYIF PVHWQFGQLD QHPIDGYLSH TELAPLRAPL IPMEHCTTRF FETCDLDNDK YIALDEWAGC FGIKQKDIDK DLVI (1-303 aa encoded by BC004974)
SPARC gene encodes a cysteine-rich acidic matrix-associated protein. The encoded protein is required for the collagen in bone to become calcified but is also involved in extracellular matrix synthesis and promotion of changes to cell shape. The gene product has been associated with tumor suppression but has also been correlated with metastasis based on changes to cell shape which can promote tumor cell invasion. Three transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Basement membrane protein 40; basement-membrane protein 40; BM-40; BM40; Cysteine rich protein; ON; osteonectin; Secreted acidic cystein rich glycoprotein; Secreted acidic cystein-rich glycoprotein (osteonectin); secreted acidic cysteine rich glycoprotein; secreted protein acidic and rich in cysteine; Secreted protein acidic and rich in cysteine (SPARC); secreted protein, acidic, cysteine-rich (osteonectin); SPARC; unnamed protein product
Gene Aliases: BM-40; OI17; ON; ONT
UniProt ID: (Human) P09486, (Pig) P20112, (Mouse) P07214, (Rat) P16975
Entrez Gene ID: (Human) 6678, (Pig) 595124, (Mouse) 20692, (Rat) 24791
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support