Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • SRPX2 Antibodies

          Proteintech

          SRPX2 Polyclonal Antibody

          7 References
          View all (14) SRPX2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite SRPX2 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (21)
          SRPX2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          SRPX2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 21

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          SRPX2 Antibody (11845-1-AP) in IHC (P)

          Immunohistochemistry of paraffin-embedded human placenta using 11845-1-AP (SRPX2 antibody) at dilution of 1:100 (under 40x lens). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          SRPX2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SRPX2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SRPX2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SRPX2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SRPX2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SRPX2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SRPX2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SRPX2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SRPX2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SRPX2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SRPX2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SRPX2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SRPX2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SRPX2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SRPX2 Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
          SRPX2 Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
          SRPX2 Antibody in Western Blot (WB)
          SRPX2 Antibody in Western Blot (WB)
          SRPX2 Antibody in Western Blot (WB)
          SRPX2 Antibody in Western Blot (WB)
          SRPX2 Antibody in Western Blot (WB)
          SRPX2 Polyclonal Antibody

          Product Details

          11845-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          View 5 publications 5 publications

          Immunohistochemistry (IHC)

          -
          View 2 publications 2 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20-1:200
          -

          Immunohistochemistry (PFA fixed) (IHC (PFA))

          1:100
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          SRPX2 Fusion Protein Ag2516 (166-465 aa encoded by BC020733)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.14 mg/mL

          Amount

          21 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: DGRWSGGEP VCVDIDPPKI RCPHSREKMA EPEKLTARVY WDPPLVKDSA DGTITRVTLR GPEPGSHFPE GEHVIRYTAY DRAYNRASCK FIVKVQVRRC PTLKPPQHGY LTCTSAGDNY GASCEYHCDG GYDRQGTPSR VCQSSRQWSG SPPICAPMKI NVNVNSAAGL LDQFYEKQRL LIISAPDPSN RYYKMQISML QQSTCGLDLR HVTIIELVGQ PPQEVGRIRE QQLSANIIEE LRQFQRLTRS YFNMVLIDKQ GIDRDRYMEP VTPEEIFTFI DDYLLSNQEL TQRREQRDIC E (166-465 aa encoded by BC020733)

          Target Information

          Sushi-repeat-containing protein X-linked 2 (SRPX2) is a neural gene functioning in the speech and language center of the human brain; mutations in this gene lead to epilepsy, speech dyspraxia, mental retardation and cognitive disorders. Recently, SRPX2 was found to be a novel mediator of angiogenesis and can act as a ligand for the urokinase-type plasminogen activator, a protein that can facilitate invasive migration of sprouting endothelial cells. SRPX2 is also overexpressed in gastric cancer, leading to increased phosphorylation levels of focal adhesion kinase and enhanced cellular migration and adhesion, suggesting that SRPX2 may be a potential target in the treatment of metastatic cancers.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: RP11-524D16__A.1; SRP; SRPUL; srpx2 {ECO:0000312|EMBL:AAI68975.1, ECO:0000312|RGD:1562444}; Sushi repeat-containing protein SRPX2; sushi repeat-containing protein SRPX2 {ECO:0000303|PubMed:24179158}; sushi-repeat containing protein; sushi-repeat protein up-regulated in leukemia; Sushi-repeat protein upregulated in leukemia; unnamed protein product

          View more View less

          Gene Aliases: 1110039C07Rik; BPP; CBPS; PMGX; RESDX; RGD1562444; SRP; SRPUL; SRPX2

          View more View less

          UniProt ID: (Human) O60687, (Rat) B5DF94, (Mouse) Q8R054

          View more View less

          Entrez Gene ID: (Human) 27286, (Rat) 317181, (Mouse) 68792

          View more View less

          Function(s)
          receptor binding extracellular matrix structural constituent protein binding hepatocyte growth factor binding identical protein binding
          Process(es)
          angiogenesis cell adhesion regulation of phosphorylation cell motility regulation of synapse assembly positive regulation of synapse assembly vocalization behavior positive regulation of cell migration involved in sprouting angiogenesis cell-cell adhesion single organismal cell-cell adhesion
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-kg5vd:80/100.66.78.64:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0