Search
Search
Bioss
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
The protein encoded by this gene catalyzes the conversion of sulfated steroid precursors to estrogens during pregnancy. The encoded protein is found in the endoplasmic reticulum, where it acts as a homodimer. Mutations in this gene are known to cause X-linked ichthyosis (XLI).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Arylsulfatase C; ASC; Estrone sulfatase; Steroid sulfatase; steroid sulfatase (microsomal), isozyme S; steroid sulfatase precursor (EC 3.1.6.2); steroidogenic enzyme; Steryl-sulfatase; Steryl-sulfate sulfohydrolase; unnamed protein product
Gene Aliases: ARSC; ARSC1; ASC; ES; SSDD; STS; XLI
UniProt ID: (Human) P08842, (Mouse) P50427, (Rat) P15589
Entrez Gene ID: (Human) 412, (Mouse) 20905, (Rat) 24800
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support