Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Synaptojanin 2 Antibodies

          Abnova

          SYNJ2 Polyclonal Antibody, MaxPab™

          View all (14) Synaptojanin 2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite SYNJ2 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          SYNJ2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          SYNJ2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          SYNJ2 Antibody (H00008871-D01P) in WB

          Western Blot analysis of SYNJ2 expression in transfected 293T cell line (H00008871-T02) by SYNJ2 MaxPab polyclonal antibody.

          Lane 1: SYNJ2 transfected lysate(92.20 KDa).
          Lane 2: Non-transfected lysate.
          {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          SYNJ2 Antibody in Western Blot (WB)
          SYNJ2 Polyclonal Antibody, MaxPab™

          Product Details

          H00008871-D01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1-5 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          SYNJ2 (AAH43277.1, 1 a.a. approximately 839 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          See Label

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Sequence of this protein is as follows: MGGKAGNKGA VGIRFQFHST SFCFICSHLT AGQSQVKERN EDYKEITQKL CFPMGRNVFS HDYVFWCGDF NYRIDLTYEE VFYFVKRQDW KKLLEFDQLQ LQKSSGKIFK DFHEGAINFG PTYKYDVGSA AYDTSDKCRT PAWTDRVLWW RKKHPFDKTA GELNLLDSDL DVDTKVRHTW SPGALQYYGR AELQASDHRP VLAIVEVEVQ EVDVGARERV FQEVSSFQGP LDATVVVNLQ SPTLEEKNEF PEDLRTELMQ TLGSYGTIVL VRINQGQMLV TFADSHSALS VLDVDGMKVK GRAVKIRPKT KDWLKGLREE IIRKRDSMAP VSPTANSCLL EENFDFTSLD YESEGDILED DEDYLVDEFN QPGVSDSELG GDDLSDVPGP TALAPPSKSP ALTKKKQHPT YKDDADLVEL KRELEAVGEF RHRSPSRSLS VPNRPRPPQP PQRPPPPTGL MVKKSASDAS ISSGTHGQYS ILQTARLLPG APQQPPKART GISKPYNVKQ IKTTNAQEAE AAIRCLLEAR GGASEEALSA VAPRDLEASS EPEPTPGAAK PETPQAPPLL PRRPPPRVPA IKKPTLRRTG KPLSPEEQFE QQTVHFTIGP PETSVEAPPV VTAPRVPPVP KPRTFQPGKA AERPSHRKPA SDEAPPGAGA SVPPPLEAPP LVPKVPPRRK KSAPAAFHLQ VLQSNSQLLQ GLTYNSSDSP SGHPPAAGTV FPQGDFLSTS SATSPDSDGT KAMKPEAAPL LGDYQDPFWN LLHHPKLLNN TWLSKSSDPL DSGTRSPKRD PIDPVSAGAS AAKAELPPDH GHKTLGHWVT ISDQEKRTAL QVFDPLAKT

          Target Information

          The gene is a member of the inositol-polyphosphate 5-phosphatase family. The encoded protein interacts with the ras-related C3 botulinum toxin substrate 1, which causes translocation of the encoded protein to the plasma membrane where it inhibits clathrin-mediated endocytosis. Alternative splicing results in multiple transcript variants.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: inositol phosphate 5'-phosphatase 2; inositol polyphosphate 5'-phosphatase; inositol polyphosphate-5-phosphatase H; phosphoinositide 5-phosphatase; Synaptic inositol 1,4,5-trisphosphate 5-phosphatase 2; Synaptojanin-2; unnamed protein product

          View more View less

          Gene Aliases: INPP5H; KIAA0348; SYNJ2

          View more View less

          UniProt ID: (Human) O15056

          View more View less

          Entrez Gene ID: (Human) 8871

          View more View less

          Function(s)
          nucleic acid binding RNA binding phosphatidylinositol-3-phosphatase activity phosphatidylinositol-4,5-bisphosphate 5-phosphatase activity protein binding hydrolase activity phosphatase activity phosphatidylinositol phosphate 4-phosphatase activity phosphatidylinositol-3,5-bisphosphate 5-phosphatase activity phosphatidylinositol-3,5-bisphosphate 3-phosphatase activity inositol-1,4,5-trisphosphate 5-phosphatase activity phosphatidylinositol phosphate phosphatase activity
          Process(es)
          lipid metabolic process phosphatidylinositol biosynthetic process phosphatidylinositol-3-phosphate biosynthetic process phosphatidylinositol dephosphorylation synaptic vesicle endocytosis membrane organization
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-lcddp:80/100.66.76.150:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0