Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • SIRT6 Antibodies

          Bethyl Laboratories

          Sirt6 Polyclonal Antibody

          View all (30) SIRT6 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Sirt6 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          Sirt6 Antibody in Immunoprecipitation (IP)
          Group 53 Created with Sketch.
          Sirt6 Antibody in Immunoprecipitation (IP)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Sirt6 Antibody (A302-451A) in IP

          Detection of human Sirt6 by western blot of immunoprecipitates. Samples: Whole cell lysate (1 mg for IP, 20% of IP loaded) from HeLa cells. Antibodies: Affinity purified rabbit anti-Sirt6 antibody (Product # A302-451A) used for IP at 3 µg/mg lysate. Sirt6 was also immunoprecipitated by rabbit anti-Sirt6 antibody A302-452A, which recognizes a downstream epitope. For blotting immunoprecipitated Sirt6, A302-452A was used at 1 µg/mL. Detection: Chemiluminescence with an exposure time of 30 seconds. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Sirt6 Antibody in Immunoprecipitation (IP)
          Sirt6 Polyclonal Antibody

          Product Details

          A302-451A

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          Assay-dependent
          -

          Immunoprecipitation (IP)

          2-5 µg/mg lysate
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Region between residue 50 and 100 of Human sirtuin (silent mating type information regulation 2 homolog) 6.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          tris citrate/phosphate, pH 7-8

          Contains

          0.09% sodium azide

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          The recommended shelf life for this product is 1 year from date of receipt.

          Based on 100% sequence identity, this antibody is predicted to react with Mouse

          Target Information

          Sirtuins regulate epigenetic gene silencing. SIRT6 (Mono-ADP-ribosyltransferase sirtuin-6), a mouse Sir2 homolog, is a chromatin-associated nuclear ADP-ribosyltransferase protein. It appears to be involved in DNA repair and may also play a role in human aging.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: hSIRT6; NAD-dependent protein deacetylase sirtuin-6; NAD-dependent protein deacylase sirtuin-6; Protein mono-ADP-ribosyltransferase sirtuin-6; Regulatory protein SIR2 homolog 6; SIR2; SIR2 like 6; SIR2-like protein 6; sir2-related protein type 6; SIRT6; sirtuin type 6; sirtuin6; unnamed protein product

          View more View less

          Gene Aliases: hSIRT6; SIR2L6; SIRT6

          View more View less

          UniProt ID: (Human) Q8N6T7

          View more View less

          Entrez Gene ID: (Human) 51548

          View more View less

          Function(s)
          DNA binding chromatin binding damaged DNA binding transcription corepressor activity RNA binding NAD+ ADP-ribosyltransferase activity protein binding zinc ion binding transferase activity transferase activity, transferring acyl groups transferase activity, transferring glycosyl groups nucleotidyltransferase activity NAD-dependent histone deacetylase activity enzyme regulator activity chromatin DNA binding nucleosome binding histone deacetylase activity (H3-K9 specific) NAD-dependent protein deacetylase activity histone deacetylase regulator activity protein homodimerization activity metal ion binding NAD-dependent histone deacetylase activity (H3-K9 specific) NAD+ binding NAD-dependent histone deacetylase activity (H3-K18 specific) NAD+-protein-arginine ADP-ribosyltransferase activity DNA damage sensor activity histone H3K56 deacetylase activity, NAD-dependent NAD-dependent protein demyristoylase activity NAD-dependent protein depalmitoylase activity NAD+-protein-lysine ADP-ribosyltransferase activity TORC2 complex binding protein ADP-ribosylase activity
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter gluconeogenesis glycolytic process regulation of gluconeogenesis DNA repair regulation of DNA repair base-excision repair double-strand break repair chromatin organization chromatin remodeling protein deacetylation protein import into nucleus inflammatory response cellular response to DNA damage stimulus Ras protein signal transduction negative regulation of cell proliferation determination of adult lifespan response to UV negative regulation of transposition, RNA-mediated regulation of double-strand break repair via homologous recombination regulation of lipid metabolic process pericentric heterochromatin assembly telomeric heterochromatin assembly protein destabilization positive regulation of insulin secretion positive regulation of telomere maintenance positive regulation of proteasomal ubiquitin-dependent protein catabolic process circadian regulation of gene expression negative regulation of transcription elongation from RNA polymerase II promoter ketone biosynthetic process negative regulation of protein import into nucleus glucose homeostasis regulation of circadian rhythm positive regulation of fat cell differentiation negative regulation of gluconeogenesis positive regulation of gluconeogenesis negative regulation of gene expression, epigenetic negative regulation of glycolytic process negative regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter negative regulation of glucose import positive regulation of protein export from nucleus positive regulation of fibroblast proliferation regulation of protein secretion regulation of lipid catabolic process protein delipidation cardiac muscle cell differentiation protein poly-ADP-ribosylation positive regulation of cold-induced thermogenesis negative regulation of protein localization to chromatin positive regulation of protein localization to chromatin DNA repair-dependent chromatin remodeling transcription pausing by RNA polymerase II positive regulation of stem cell population maintenance positive regulation of chondrocyte proliferation regulation of protein localization to plasma membrane positive regulation of blood vessel branching positive regulation of vascular endothelial cell proliferation protein localization to site of double-strand break positive regulation of stem cell proliferation positive regulation of stem cell differentiation negative regulation of cellular senescence positive regulation of cellular senescence positive regulation of double-strand break repair
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline