Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
This antibody recognizes the mature approximately 75-100 kDa protein and a partially glycosylated 50-55 kDa protein.
Immunogen sequence: TVLCGALIS CLTGPTKRST LAPGLLWWDL ARQTASVAPK EEVAILDDNL VKGPEELPTG NKKPPGFLPT NEDRLFFLGQ KELEGAGSWT PCVGHDGGRD QQETNL (539-643 aa encoded by BC105047)
Human Sodium Iodide Symporter (hNIS) is responsible for iodide concentrating ability within thyroid follicular cells. It is a membrane bound glycoprotein with 13 membrane spanning domains and 14 extramembranous domains. It may represent an autoantigen in thyroid.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Na(+)/I(-) cotransporter; Na(+)/I(-) symporter; Na+/I- symporter; natrium iodide transporter; sodium-iodide symporter; sodium/iodide symporter; solute carrier family 5 (sodium iodide symporter), member 5; solute carrier family 5 (sodium/iodide cotransporter), member 5; solute carrier family 5 member 5; symporter; NIS
Gene Aliases: NIS; TDH1
UniProt ID: (Human) Q92911, (Rat) Q63008
Entrez Gene ID: (Human) 6528, (Mouse) 114479, (Rat) 114613
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support