Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 3
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
This antibody is predicted to react with canine based on sequence homology.
This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
Spinesin, also known as Serine protease 22 (PRSS22), is a gene located on chromosome 16p11.2 that encodes a trypsin-like serine protease predominantly expressed in the brain and airway tissues. This protease is part of the large family of serine proteases, characterized by their involvement in a variety of physiological processes, including digestion, immune response, and cellular remodeling. In the nervous system, Spinesin is implicated in synaptic function and plasticity, with potential roles in modulating neural connectivity and communication. It is also involved in processing extracellular matrix components, which can influence cell adhesion and migration, contributing to tissue remodeling and repair processes. Dysregulation of Spinesin expression or activity may impact neural development and has been associated with certain neurological disorders. Research into Spinesin continues to explore its specific functions in the nervous system and its potential therapeutic applications in neurobiology and regenerative medicine.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC141886; MGC148044; transmembrane protease, serine 5; unnamed protein product
Gene Aliases: SPINESIN
UniProt ID: (Human) Q9H3S3
Entrez Gene ID: (Human) 80975
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support