    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • HCFC2 Antibodies
          OriGene
          HCFC2 Monoclonal Antibody (OTI5H9), TrueMAB™
          View all (13) HCFC2 antibodies
          Questions & Answers
          Cite HCFC2 Monoclonal Antibody (OTI5H9), TrueMAB™
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (6)
          HCFC2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          HCFC2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 6
          HCFC2 Antibody (TA505235S) in ICC/IF
          Anti-HCFC2 mouse monoclonal antibody (TA505235) immunofluorescent staining of COS7 cells transiently transfected by pCMV6-ENTRY HCFC2(RC207543). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          HCFC2 Antibody in Immunocytochemistry (ICC/IF)
          HCFC2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HCFC2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HCFC2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HCFC2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HCFC2 Antibody in Western Blot (WB)
          HCFC2 Monoclonal Antibody (OTI5H9), TrueMAB™
          Product Details
          TA505235S
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:2,000
          -
          Immunohistochemistry (Paraffin) (IHC (P))
          1:150
          -
          Immunocytochemistry (ICC/IF)
          1:100
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          OTI5H9
          Immunogen
          Full length human recombinant protein of human HCFC2 produced in HEK293T cell.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          30 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS with 1% BSA, 50% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          This gene encodes one of two proteins which interact with VP16, a herpes simplex virus protein that initiates virus infection. Both the encoded protein and the original Herpes host cell factor interact with VP16 through a beta-propeller domain. The original Herpes host cell factor, however, is effective at initiating viral infection while the encoded protein is not. Transcripts of varying length due to alternative polyadenylation signals have been described.
          Bioinformatics
          Protein Aliases: C2 factor; HCF-2; Host cell factor 2; unnamed protein product
          View more View less
          Gene Aliases: HCF-2; HCF2; HCFC2
          View more View less
          UniProt ID: (Human) Q9Y5Z7
          View more View less
          Entrez Gene ID: (Human) 29915
          View more View less
          Function(s)
          transcription cofactor activity transcription coactivator activity protein binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter chromatin remodeling regulation of transcription, DNA-templated regulation of transcription from RNA polymerase II promoter viral process regulation of toll-like receptor 3 signaling pathway positive regulation of transcription, DNA-templated antiviral innate immune response immune response involved in response to exogenous dsRNA
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline