    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CD1c Antibodies
          OriGene
          CD1C Monoclonal Antibody (OTI2D4), Biotin, TrueMAB™
          View all (84) CD1c antibodies
          Questions & Answers
          Cite CD1C Monoclonal Antibody (OTI2D4), Biotin, TrueMAB™
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          CD1C Antibody in ELISA (ELISA)
          Group 53 Created with Sketch.
          CD1C Antibody in ELISA (ELISA)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          CD1C Antibody (TA700316) in ELISA
          CD1C Elisa with 4C7 Capture (TA600316) and 2D4 Detection (TA700316) Antibodies. Substrate used: Recombinant Human CD1C (TP318490) {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          CD1C Antibody in ELISA (ELISA)
          CD1C Antibody in Luminex (Luminex)
          CD1C Monoclonal Antibody (OTI2D4), Biotin, TrueMAB™
          Product Details
          TA700316
          Applications
          Tested Dilution
          Publications
          ELISA (ELISA)
          1:250-1:1,000
          -
          Luminex (Luminex)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          OTI2D4
          Immunogen
          Full length human recombinant protein of human CD1C produced in HEK293T cell.
          3D Epitope / Immunogen
          Conjugate
          Biotin Biotin Biotin
          • Unconjugated
          • Request custom conjugation
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          50 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS with 10mg/mL BSA, 50% glycerol
          Contains
          0.05% sodium azide
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          CD1c is a member of the CD1 family of transmembrane glycoproteins, which are structurally related to major histocompatibility complex (MHC) proteins and form heterodimers with beta-2-microglobulin. This family of proteins is involved in the presentation of lipid and glycolipid antigens, both of self and microbial origin, to T cells during the adaptive immune response. CD1c is expressed on some circulating and marginal zone B cells, as well as in lymph nodes and germinal centers. It plays a crucial role in presenting lipid antigens, such as microbial fatty acids, to effector T cells. The protein encoded by the CD1c gene localizes to late endosomes and lysosomes, utilizing a tyrosine-based motif in its cytoplasmic tail for targeting. Vesicular acidification is required for CD1c to bind lipid antigens effectively. The human genome contains five CD1 family genes organized in a cluster on chromosome 1, with each member differing in cellular localization and specificity for particular lipid ligands. CD1c undergoes alternative splicing, resulting in three different isoforms: soluble, membrane-bound, and cytoplasmic/soluble isoforms, highlighting its functional diversity in immune processes.
          Bioinformatics
          Protein Aliases: CD1C; CD1c antigen precursor; CD1C antigen, c polypeptide; cortical thymocyte antigen CD1C; differentiation antigen CD1-alpha-3; RP11-101J8.3; T-cell surface glycoprotein CD1c; unnamed protein product
          View more View less
          Gene Aliases: BDCA1; CD1; CD1A; CD1C; R7
          View more View less
          UniProt ID: (Human) P29017
          View more View less
          Entrez Gene ID: (Human) 911
          View more View less
          Function(s)
          protein binding lipid binding endogenous lipid antigen binding exogenous lipid antigen binding glycolipid binding lipopeptide binding
          Process(es)
          positive regulation of T cell mediated cytotoxicity adaptive immune response T cell activation involved in immune response immune system process immune response antigen processing and presentation, endogenous lipid antigen via MHC class Ib antigen processing and presentation, exogenous lipid antigen via MHC class Ib
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline