Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • TCF7L2 Antibodies

          Abnova

          TCF7L2 Polyclonal Antibody, MaxPab™

          View all (24) TCF7L2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite TCF7L2 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          TCF7L2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          TCF7L2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          TCF7L2 Antibody (H00006934-B01P) in WB

          Western Blot analysis of TCF7L2 expression in transfected 293T cell line by TCF7L2 MaxPab polyclonal antibody. Lane 1: TCF7L2 transfected lysate(50.60 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          TCF7L2 Antibody in Western Blot (WB)
          TCF7L2 Polyclonal Antibody, MaxPab™

          Product Details

          H00006934-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          TCF7L2 (AAH32656.1, 1 a.a. approximately 455 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5-1 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MPQLNGGGGD DLGANDELIS FKDEGEQEEK SSENSSAERD LADVKSSLVN ESETNQNSSS DSEAERRPPP RSESFRDKSR ESLEEAAKRQ DGGLFKGPPY PGYPFIMIPD LTSPYLPNGS LSPTARTYLQ MKWPLLDVQA GSLQSRQALK DARSPSPAHI VSNKVPVVQH PHHVHPLTPL ITYSNEHFTP GNPPPHLPAD VDPKTGIPRP PHPPDISPYY PLSPGTVGQI PHPLGWQGQP VYPITTGGFR HPYPTALTVN ASMSRFPPHM VPPHHTLHTT GIPHPAIVTP TVKQESSQSD VGSLHSSKHQ DSKKEEEKKK PHIKKPLNAF MLYMKEMRAK VVAECTLKES AAINQILGRR WHALSREEQA KYYELARKER QLHMQLYPGW SARDNYGKKK KRKRDKQPGE TNEHSECFLN PCLSLPPITG EKKSAFATYK VKAAASAHPL QMEAY

          Target Information

          TCF7L2, or TCF4, is a high mobility group (HMG) box-containing transcription factor that plays a key role in the Wnt signaling pathway. TCF7L2 has been implicated in glucose homeostasis through the regulation of pro-glucagon gene expression, which encodes glucagon-like peptide 1 (GLP-1). Genetic variants of TCF7L2 have been linked to type 2 diabetes, and it has specifically been shown to have an important role in the regulation of both beta-cell survival and function. Several splicing isoforms for TCF7L2 have been shown to be differentially expressed in tissues during cancer progression, and gene targeting studies indicate that TCF7L2 is required to maintain the crypt stem cells of the small intestine.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: contains exons 1, 2, 3, 4, 5, 6, 7L, 8, 9L, 10, 11, 12, 13, 17; contains exons 1, 2, 3, 4, 5, 6, 7L, 8, 9L, 10, 11, 12, 15, 17; contains exons 1, 2, 3, 4, 5, 6, 7L, 8, 9L, 10, 11, 12, 17; contains exons 1, 2, 3, 4, 5, 6, 7L, 8, 9S, 10, 11, 12, 13, 14, 17; contains exons 1, 2, 3, 4, 5, 6, 7L, 8, 9S, 10, 11, 12, 13, 17; contains exons 1, 2, 3, 4, 5, 6, 7L, 8, 9S, 10, 11, 12, 17; contains exons 1, 2, 3, 4, 5, 6, 7S, 8, 9S, 10, 11, 12, 17; contains exons 1, 2, 3, 5, 6, 7L, 8, 9L, 10, 11, 12, 15, 17; contains exons 1, 2, 3, 5, 6, 7L, 8, 9L, 10, 11, 12, 17; contains exons 1, 2, 3, 5, 6, 7L, 8, 9S, 10, 11, 12, 13, 14, 17; contains exons 1, 2, 3, 5, 6, 7L, 8, 9S, 10, 11, 12, 15, 17; contains exons 1, 2, 3, 5, 6, 7L, 8, 9S, 10, 11, 12, 17; contains exons 1, 2, 3, 5, 6, 7S, 8, 9S, 10, 11, 12, 17; contains exons 8, 9L, 10, 11, 12, 13, 14, 17; HMG box transcription factor 4; hTCF; hTCF 4; hTCF-4; RP11-357H24.1; T-cell factor 4; T-cell-specific transcription factor 4; TCF 4; TCF7L2; TCF7L2 protein; transcription factor; Transcription factor 7-like 2; transcription factor 7-like 2 (T-cell specific, HMG-box); unnamed protein product

          View more View less

          Gene Aliases: TCF-4; TCF4; TCF7L2

          View more View less

          UniProt ID: (Human) Q9NQB0

          View more View less

          Entrez Gene ID: (Human) 6934

          View more View less

          Function(s)
          transcription regulatory region sequence-specific DNA binding RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding transcription corepressor binding transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding DNA binding transcription factor activity, sequence-specific DNA binding protein binding beta-catenin binding ligand-dependent nuclear receptor binding protein kinase binding sequence-specific DNA binding gamma-catenin binding RNA polymerase II sequence-specific DNA binding transcription factor binding armadillo repeat domain binding promoter-specific chromatin binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter blood vessel development regulation of transcription from RNA polymerase II promoter response to glucose positive regulation of epithelial to mesenchymal transition positive regulation of heparan sulfate proteoglycan biosynthetic process Wnt signaling pathway pancreas development positive regulation of insulin secretion regulation of hormone metabolic process glucose homeostasis maintenance of DNA repeat elements fat cell differentiation negative regulation of gluconeogenesis negative regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter myoblast fate commitment regulation of smooth muscle cell proliferation positive regulation of epithelial cell proliferation positive regulation of protein kinase B signaling canonical Wnt signaling pathway negative regulation of androgen receptor signaling pathway negative regulation of canonical Wnt signaling pathway positive regulation of protein localization to nucleus negative regulation of type B pancreatic cell apoptotic process negative regulation of extrinsic apoptotic signaling pathway
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-lcddp:80/100.66.76.150:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0