Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • TCF7L2 Antibodies

          Invitrogen

          TCF7L2 Polyclonal Antibody

          Advanced Verification
          View all (24) TCF7L2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite TCF7L2 Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (5)
          • Advanced Verification (1)
          TCF7L2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          TCF7L2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 6

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          TCF7L2 Antibody (PA5-78192) in ICC/IF

          TCF7L2 Polyclonal Antibody detects TCF7L2 protein at nucleus by immunofluorescent analysis. Sample: HeLa cells were fixed in 4% paraformaldehyde at RT for 15 min. Green: TCF7L2 protein stained by TCF7L2 Polyclonal Antibody (Product # PA5-78192) diluted at 1:500. Red: Phalloidin, a cytoskeleton marker, diluted at 1:100. Scale bar = 10 µm. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          TCF7L2 Antibody in Immunocytochemistry (ICC/IF)
          TCF7L2 Antibody in Western Blot (WB)
          TCF7L2 Antibody in Western Blot (WB)
          TCF7L2 Antibody in Western Blot (WB)
          TCF7L2 Antibody in Western Blot (WB)
          TCF7L2 Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          TCF7L2 Polyclonal Antibody

          Product Details

          PA5-78192

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:3,000
          -

          Immunocytochemistry (ICC/IF)

          1:100-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human TCF7L2. The exact sequence is proprietary.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1.91 mg/mL

          Amount

          191 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7, with 20% glycerol

          Contains

          0.025% ProClin 300

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2736692

          Product Specific Information

          Positive Control: 293T, A431, HeLa, HepG2

          Predicted Reactivity: Mouse (100%), Rat (100%), Zebrafish (100%), Bovine (100%)

          Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

          Target Information

          TCF7L2, or TCF4, is a high mobility group (HMG) box-containing transcription factor that plays a key role in the Wnt signaling pathway. TCF7L2 has been implicated in glucose homeostasis through the regulation of pro-glucagon gene expression, which encodes glucagon-like peptide 1 (GLP-1). Genetic variants of TCF7L2 have been linked to type 2 diabetes, and it has specifically been shown to have an important role in the regulation of both beta-cell survival and function. Several splicing isoforms for TCF7L2 have been shown to be differentially expressed in tissues during cancer progression, and gene targeting studies indicate that TCF7L2 is required to maintain the crypt stem cells of the small intestine.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: alternatively spliced; short isoform; lacks DNA binding domain; contains exons 1, 2, 3, 4, 5, 6, 7L, 8, 9L, 10, 11, 12, 13, 17; contains exons 1, 2, 3, 4, 5, 6, 7L, 8, 9L, 10, 11, 12, 15, 17; contains exons 1, 2, 3, 4, 5, 6, 7L, 8, 9L, 10, 11, 12, 17; contains exons 1, 2, 3, 4, 5, 6, 7L, 8, 9S, 10, 11, 12, 13, 14, 17; contains exons 1, 2, 3, 4, 5, 6, 7L, 8, 9S, 10, 11, 12, 13, 17; contains exons 1, 2, 3, 4, 5, 6, 7L, 8, 9S, 10, 11, 12, 17; contains exons 1, 2, 3, 4, 5, 6, 7S, 8, 9S, 10, 11, 12, 17; contains exons 1, 2, 3, 5, 6, 7L, 8, 9L, 10, 11, 12, 15, 17; contains exons 1, 2, 3, 5, 6, 7L, 8, 9L, 10, 11, 12, 17; contains exons 1, 2, 3, 5, 6, 7L, 8, 9S, 10, 11, 12, 13, 14, 17; contains exons 1, 2, 3, 5, 6, 7L, 8, 9S, 10, 11, 12, 15, 17; contains exons 1, 2, 3, 5, 6, 7L, 8, 9S, 10, 11, 12, 17; contains exons 1, 2, 3, 5, 6, 7S, 8, 9S, 10, 11, 12, 17; contains exons 8, 9L, 10, 11, 12, 13, 14, 17; HMG box transcription factor 4; HMG box transcription factor TCF7L2; hTCF; hTCF 4; hTCF-4; RP11-357H24.1; T-cell factor 4; T-cell-specific transcription factor 4; TCF 4; TCF7L2; TCF7L2 protein; transcription factor; transcription factor 7-like 2 (T-cell specific, HMG-box); unnamed protein product

          View more View less

          Gene Aliases: TCF-4; TCF4; TCF4B; TCF4E

          View more View less

          UniProt ID: (Human) Q9NQB0, (Mouse) Q924A0

          View more View less

          Entrez Gene ID: (Human) 6934, (Rat) 679869, (Mouse) 21416

          View more View less

          Function(s)
          transcription regulatory region sequence-specific DNA binding RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding transcription corepressor binding transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding transcription factor activity, sequence-specific DNA binding protein binding beta-catenin binding ligand-dependent nuclear receptor binding protein kinase binding sequence-specific DNA binding gamma-catenin binding RNA polymerase II sequence-specific DNA binding transcription factor binding armadillo repeat domain binding promoter-specific chromatin binding chromatin binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter blood vessel development regulation of gluconeogenesis regulation of transcription from RNA polymerase II promoter response to glucose positive regulation of epithelial to mesenchymal transition positive regulation of heparan sulfate proteoglycan biosynthetic process Wnt signaling pathway pancreas development positive regulation of insulin secretion regulation of hormone metabolic process glucose homeostasis maintenance of DNA repeat elements fat cell differentiation negative regulation of gluconeogenesis negative regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter myoblast fate commitment regulation of smooth muscle cell proliferation positive regulation of epithelial cell proliferation positive regulation of protein kinase B signaling canonical Wnt signaling pathway negative regulation of androgen receptor signaling pathway negative regulation of canonical Wnt signaling pathway positive regulation of protein localization to nucleus negative regulation of type B pancreatic cell apoptotic process negative regulation of extrinsic apoptotic signaling pathway cellular glucose homeostasis in utero embryonic development glycogen metabolic process glucose metabolic process regulation of transcription, DNA-templated cellular response to starvation post-embryonic development tissue development positive regulation of triglyceride biosynthetic process oligodendrocyte development neural tube development pituitary gland development cell differentiation bone mineralization negative regulation of BMP signaling pathway embryonic genitalia morphogenesis regulation of myelination secretory granule localization somatic stem cell population maintenance multicellular organism growth negative regulation of fibroblast growth factor receptor signaling pathway odontogenesis of dentin-containing tooth positive regulation of apoptotic process skin development negative regulation of fat cell differentiation positive regulation of gluconeogenesis positive regulation of transcription, DNA-templated negative regulation of organ growth positive regulation of lipid biosynthetic process embryonic digestive tract morphogenesis embryonic hindgut morphogenesis regulation of skeletal muscle tissue development regulation of oligodendrocyte differentiation face morphogenesis regulation of insulin secretion involved in cellular response to glucose stimulus insulin metabolic process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-74676f44b6-nqctq:80/100.66.72.150:80.
          git-commit: db542085ed6beb0c3f0dd774c2548ab49b9f1b6a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026.10.42-1.0