Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • TDRD1 Antibodies

          Abnova

          TDRD1 Polyclonal Antibody, MaxPab™

          View all (10) TDRD1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite TDRD1 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          TDRD1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          TDRD1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          TDRD1 Antibody (H00056165-B01P) in WB

          Western Blot analysis of TDRD1 expression in transfected 293T cell line by TDRD1 MaxPab polyclonal antibody. Lane 1: TDRD1 transfected lysate(130.79 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          TDRD1 Antibody in Western Blot (WB)
          TDRD1 Polyclonal Antibody, MaxPab™

          Product Details

          H00056165-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          TDRD1 (NP_942090.1, 1 a.a. approximately 1189 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MSVKSPFNVM SRNNLEAPPC KMTEPFNFEK NENKLPPHES LRSPGTLPNH PNFRLKSSEN GNKKNNFLLC EQTKQYLASQ EDNSVSSNPN GINGEVVGSK GDRKKLPAGN SVSPPSAESN SPPKEVNIKP GNNVRPAKSK KLNKLVENSL SISNPGLFTS LGPPLRSTTC HRCGLFGSLR CSQCKQTYYC STACQRRDWS AHSIVCRPVQ PNFHKLENKS SIETKDVEVN NKSDCPLGVT KEIAIWAERI MFSDLRSLQL KKTMEIKGTV TEFKHPGDFY VQLYSSEVLE YMNQLSASLK ETYANVHEKD YIPVKGEVCI AKYTVDQTWN RAIIQNVDVQ QKKAHVLYID YGNEEIIPLN RIYHLNRNID LFPPCAIKCF VANVIPAEGN WSSDCIKATK PLLMEQYCSI KIVDILEEEV VTFAVEVELP NSGKLLDHVL IEMGYGLKPS GQDSKKENAD QSDPEDVGKM TTENNIVVDK SDLIPKVLTL NVGDEFCGVV AHIQTPEDFF CQQLQSGRKL AELQASLSKY CDQLPPRSDF YPAIGDICCA QFSEDDQWYR ASVLAYASEE SVLVGYVDYG NFEILSLMRL CPIIPKLLEL PMQAIKCVLA GVKPSLGIWT PEAICLMKKL VQNKIITVKV VDKLENSSLV ELIDKSETPH VSVSKVLLDA GFAVGEQSMV TDKPSDVKET SVPLGVEGKV NPLEWTWVEL GVDQTVDVVV CVIYSPGEFY CHVLKEDALK KLNDLNKSLA EHCQQKLPNG FKAEIGQPCC AFFAGDGSWY RALVKEILPN GHVKVHFVDY GNIEEVTADE LRMISSTFLN LPFQGIRCQL ADIQSRNKHW SEEAITRFQM CVAGIKLQAR VVEVTENGIG VELTDLSTCY PRIISDVLID EHLVLKSASP HKDLPNDRLV NKHELQVHVQ GLQATSSAEQ WKTIELPVDK TIQANVLEII SPNLFYALPK GMPENQEKLC MLTAELLEYC NAPKSRPPYR PRIGDACCAK YTSDDFWYRA VVLGTSDTDV EVLYADYGNI ETLPLCRVQP ITSSHLALPF QIIRCSLEGL MELNGSSSQL IIMLLKNFML NQNVMLSVKG ITKNVHTVSV EKCSENGTVD VADKLVTFGL AKNITPQRQS ALNTEKMYRM NCCCTELQKQ VEKHEHILLF LLNNSTNQNK FIEMKKLLKK TASLGGKPL

          Target Information

          This gene is similar to a mouse gene that encodes a tudor domain protein. Alternatively spliced transcript variants have been described but their full length sequences have not been determined.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Cancer/testis antigen 41.1; CT41.1; FLJ21082; testicular tissue protein Li 193; Tudor domain-containing protein 1; unnamed protein product

          View more View less

          Gene Aliases: CT41.1; TDRD1

          View more View less

          UniProt ID: (Human) Q9BXT4

          View more View less

          Entrez Gene ID: (Human) 56165

          View more View less

          Function(s)
          protein binding zinc ion binding metal ion binding
          Process(es)
          germ cell development spermatogenesis cell differentiation P granule organization gene silencing by RNA piRNA metabolic process meiotic cell cycle transposable element silencing by piRNA-mediated DNA methylation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline