Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • TIS11D Antibodies

          Invitrogen

          TIS11D Polyclonal Antibody

          View all (8) TIS11D antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite TIS11D Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          TIS11D Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          TIS11D Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          TIS11D Antibody (PA5-36248) in WB

          Western blot analysis of TIS11D in Lane 1: PC12 whole cell lysate (40 µg), Lane 2: HCT116 whole cell lysate (40 µg), Lane 3: CT26 whole cell lysate (40 µg), Lane 4: A375 whole cell lysate (40 µg), Lane 5: Myla2059 whole cell lysate (40 µg). Samples were incubated with TIS11D polyclonal antibody (Product # PA5-36248) at a dilution of 1:500. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          TIS11D Antibody in Western Blot (WB)
          TIS11D Antibody in Western Blot (WB)
          TIS11D Polyclonal Antibody

          Product Details

          PA5-36248

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic peptide corresponding to amino acids 200-250 of human TIS11D
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.2, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2553439

          Product Specific Information

          This antibody detects endogenous protein at a molecular weight of 40 kDa.

          Purity is >95% by SDS-PAGE.

          Target Information

          TIS11D, also known as ZFP36LA (zinc finger protein 36, C3H type-like 2), BRF2 (butyrate response factor 2), ERF2 or RNF162C, is a 494 amino acid protein that localizes to the nucleus and contains two CSH1-type zinc fingers. Belonging to the TIS11 family of early response proteins, TIS11D is thought to function as a nuclear transcription factor that binds to 5'UUAUUUAUUU-3' core RNA sequences and may regulate growth factor-induced cellular responses. The gene encoding TIS11D maps to human chromosome 2, which houses over 1, 400 genes and comprises nearly 8% of the human genome. Harlequin icthyosis, a rare and morbid skin deformity, is associated with mutations in the ABCA12 gene, while the lipid metabolic disorder sitosterolemia is associated with defects in the ABCG5 and ABCG8 genes.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Butyrate response factor 2; butyrate response factor 2 (EGF-response factor 2); EGF-response factor 2; ERF-2; mRNA decay activator protein ZFP36L2; Protein TIS11D; TISD; TPA-induced sequence 11d; ZFP36-like 2; zinc finger protein 36, C3H type-like 1; zinc finger protein 36, C3H type-like 2; zinc finger protein 36, C3H type-like 2-like; Zinc finger protein 36, C3H1 type-like 2; zinc finger protein, C3H type, 36-like 2

          View more View less

          Gene Aliases: BRF2; ERF-2; ERF2; OOMD13; OZEMA13; RNF162C; TIS11D; ZFP36L2

          View more View less

          UniProt ID: (Human) P47974, (Mouse) P23949

          View more View less

          Entrez Gene ID: (Human) 678, (Mouse) 12193, (Rat) 298765

          View more View less

          Function(s)
          RNA binding mRNA binding protein binding zinc ion binding mRNA 3'-UTR AU-rich region binding metal ion binding DNA binding AU-rich element binding poly(A) RNA binding RNA metabolism protein
          Process(es)
          MAPK cascade nuclear-transcribed mRNA catabolic process, deadenylation-dependent decay mRNA catabolic process response to wounding hemopoiesis T cell differentiation in thymus somatic stem cell population maintenance regulation of mRNA stability cellular response to fibroblast growth factor stimulus regulation of B cell differentiation negative regulation of fat cell differentiation somatic stem cell division definitive hemopoiesis 3'-UTR-mediated mRNA destabilization ERK1 and ERK2 cascade cellular response to tumor necrosis factor cellular response to epidermal growth factor stimulus cellular response to glucocorticoid stimulus cellular response to transforming growth factor beta stimulus cellular response to granulocyte macrophage colony-stimulating factor stimulus positive regulation of nuclear-transcribed mRNA catabolic process, deadenylation-dependent decay negative regulation of mitotic cell cycle phase transition negative regulation of stem cell differentiation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline