Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • TNF alpha Antibodies

          Bioss

          TNF alpha Polyclonal Antibody, Biotin

          View all (254) TNF alpha antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite TNF alpha Polyclonal Antibody, Biotin

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          TNF alpha Polyclonal Antibody, Biotin

          Product Details

          BS-0078R-BIOTIN

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:300-1:5,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:200-1:400
          -

          Immunohistochemistry (Frozen) (IHC (F))

          1:100-1:500
          -

          ELISA (ELISA)

          1:500-1,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Pig, Rabbit, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          KLH conjugated synthetic peptide derived from human TNF alpha
          3D Epitope / Immunogen

          Conjugate

          Biotin Biotin Biotin

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          TBS, pH 7.4, with 50% glycerol, 1% BSA

          Contains

          0.03% ProClin 300

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Target Information

          TNF alpha is a multifunctional proinflammatory cytokine that belongs to the tumor necrosis factor (TNF) superfamily. This cytokine is mainly secreted by macrophage and bind to its receptors, TNFRSF1A/TNFR1 and TNFRSF1B/TNFBR. TNF alpha is involved in the regulation of immune cells, cell proliferation, differentiation, apoptosis, lipid metabolism, and coagulation. TNF alpha exists as a multimer of two, three, or five noncovalently linked units, but shows a single 17 kDa band following SDS-PAGE under non-reducing conditions. Knockout studies in mice also suggested the neuroprotective function of TNF alpha, and has been observed to causes tumor necrosis when injected into tumor-bearing mice. Other functions of TNF-alpha include its role in the immune response to bacterial, viral, parasitic and certain fungal infections, as well as its role in the necrosis of specific tumors. TNF alpha causes cytolysis or cytostasis of certain transformed cells, being synergistic with interferon-gamma in its cytotoxicity. This cytokine has been implicated in a variety of diseases, including autoimmune diseases, insulin resistance, and cancer.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: APC1 protein; Cachectin; cell apoptosis-related protein; DADB-70P7.1; TNF alpha; TNF superfamily; TNF superfamily member 2; TNF α; TNF, macrophage-derived; TNF, monocyte-derived; TNF-a; TNF-alpha; TNFα; Tumor necrosis factor; tumor necrosis factor (TNF superfamily, member 2); tumor necrosis factor alpha; tumor necrosis factor alpha precursor; tumor necrosis factor ligand 1F; Tumor necrosis factor ligand superfamily member 2; tumor necrosis factor precursor; tumor necrosis factor-alpha; tumor necrotic factor alpha; unnamed protein product

          View more View less

          Gene Aliases: DIF; IMD127; RATTNF; TNF; TNF-a; TNF-alpha; TNFA; TNFalpha; Tnfsf1a; TNFSF2; TNLG1F

          View more View less

          UniProt ID: (Pig) P23563, (Rabbit) P04924, (Human) P01375, (Rat) P16599, (Mouse) P06804

          View more View less

          Entrez Gene ID: (Pig) 397086, (Rabbit) 100009088, (Human) 7124, (Rat) 24835, (Mouse) 21926

          View more View less

          Function(s)
          protease binding identical protein binding transcription regulatory region sequence-specific DNA binding cytokine activity tumor necrosis factor receptor binding protein binding death receptor agonist activity receptor agonist activity transcription regulatory region DNA binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter activation of MAPKKK activity activation of MAPK activity microglial cell activation negative regulation of endothelial cell proliferation chronic inflammatory response to antigenic stimulus positive regulation of chronic inflammatory response to antigenic stimulus positive regulation of humoral immune response mediated by circulating immunoglobulin glucose metabolic process activation of cysteine-type endopeptidase activity involved in apoptotic process humoral immune response JNK cascade intrinsic apoptotic signaling pathway in response to DNA damage response to virus vascular endothelial growth factor production negative regulation of alkaline phosphatase activity extracellular matrix organization osteoclast differentiation sequestering of triglyceride cortical actin cytoskeleton organization positive regulation of fever generation lipopolysaccharide-mediated signaling pathway negative regulation of interleukin-6 production positive regulation of interferon-gamma production positive regulation of interleukin-6 production positive regulation of interleukin-8 production receptor biosynthetic process positive regulation of peptidyl-serine phosphorylation positive regulation of heterotypic cell-cell adhesion negative regulation of myosin-light-chain-phosphatase activity negative regulation of cysteine-type endopeptidase activity involved in apoptotic process negative regulation of protein complex disassembly positive regulation of protein complex disassembly protein kinase B signaling positive regulation of JUN kinase activity positive regulation of neuron apoptotic process negative regulation of viral genome replication positive regulation of chemokine biosynthetic process positive regulation of nitric oxide biosynthetic process regulation of fat cell differentiation negative regulation of myoblast differentiation negative regulation of osteoblast differentiation positive regulation of osteoclast differentiation negative regulation of mitotic cell cycle positive regulation of translational initiation by iron negative regulation of glucose import embryonic digestive tract development positive regulation of cytokine secretion positive regulation of phagocytosis regulation of insulin secretion positive regulation of synaptic transmission regulation of synapse organization defense response to Gram-positive bacterium leukocyte tethering or rolling negative regulation of lipid catabolic process regulation of immunoglobulin secretion positive regulation of membrane protein ectodomain proteolysis positive regulation of NF-kappaB transcription factor activity response to glucocorticoid positive regulation of hair follicle development positive regulation of protein kinase B signaling positive regulation of glial cell proliferation positive regulation of calcidiol 1-monooxygenase activity epithelial cell proliferation involved in salivary gland morphogenesis regulation of branching involved in salivary gland morphogenesis negative regulation of branching involved in lung morphogenesis positive regulation of calcineurin-NFAT signaling cascade cellular response to amino acid stimulus cellular response to nicotine cellular response to organic cyclic compound positive regulation of podosome assembly endothelial cell apoptotic process protein localization to plasma membrane necroptotic signaling pathway positive regulation of cytokine production involved in inflammatory response negative regulation of beta-amyloid clearance positive regulation of NIK/NF-kappaB signaling positive regulation of synoviocyte proliferation positive regulation of superoxide dismutase activity positive regulation of beta-amyloid formation positive regulation of protein localization to plasma membrane regulation of establishment of endothelial barrier negative regulation of bicellular tight junction assembly positive regulation of I-kappaB phosphorylation negative regulation of production of miRNAs involved in gene silencing by miRNA positive regulation of vascular smooth muscle cell proliferation positive regulation of leukocyte adhesion to arterial endothelial cell positive regulation of protein localization to cell surface positive regulation of blood microparticle formation positive regulation of chemokine (C-X-C motif) ligand 2 production positive regulation of endothelial cell apoptotic process positive regulation of estradiol secretion negative regulation of extrinsic apoptotic signaling pathway in absence of ligand protein import into nucleus, translocation MAPK cascade response to hypoxia cell activation positive regulation of cytokine production regulation of protein phosphorylation positive regulation of protein phosphorylation negative regulation of L-glutamate transport negative regulation of cytokine secretion involved in immune response skeletal muscle contraction apoptotic process obsolete transformed cell apoptotic process defense response inflammatory response signal transduction multicellular organism development negative regulation of cell proliferation extrinsic apoptotic signaling pathway via death domain receptors response to radiation response to mechanical stimulus organ morphogenesis positive regulation of gene expression negative regulation of gene expression response to activity calcium-mediated signaling positive regulation of protein complex assembly response to lipopolysaccharide positive regulation of chemokine production positive regulation of interleukin-18 production tumor necrosis factor-mediated signaling pathway regulation of cell proliferation positive regulation of NF-kappaB import into nucleus response to drug defense response to bacterium positive regulation of apoptotic process positive regulation of programmed cell death regulation of I-kappaB kinase/NF-kappaB signaling positive regulation of I-kappaB kinase/NF-kappaB signaling positive regulation of cysteine-type endopeptidase activity involved in apoptotic process positive regulation of MAP kinase activity negative regulation of growth of symbiont in host cellular extravasation positive regulation of interleukin-8 biosynthetic process regulation of osteoclast differentiation positive regulation of action potential positive regulation of mitotic nuclear division positive regulation of protein kinase activity negative regulation of transcription, DNA-templated positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter positive regulation of JNK cascade positive regulation of smooth muscle cell proliferation regulation of protein secretion positive regulation of inflammatory response leukocyte migration detection of mechanical stimulus involved in sensory perception of pain positive regulation of sequence-specific DNA binding transcription factor activity positive regulation of protein transport positive regulation of NFAT protein import into nucleus positive regulation of vitamin D biosynthetic process cellular response to lipopolysaccharide apoptotic signaling pathway extrinsic apoptotic signaling pathway response to gold nanoparticle regulation of reactive oxygen species metabolic process immune response cell proliferation positive regulation of cell proliferation positive regulation of cell adhesion macrophage activation involved in immune response regulation of immunoglobulin production positive regulation of immunoglobulin production negative regulation of cytokine production involved in immune response negative regulation of systemic arterial blood pressure JAK-STAT cascade circadian rhythm response to xenobiotic stimulus response to bacterium response to salt stress response to fructose negative regulation of heart rate positive regulation of macrophage derived foam cell differentiation negative regulation of lipid storage regulation of metabolic process positive regulation of defense response negative regulation of myelination response to nutrient levels positive regulation of fractalkine production positive regulation of interleukin-1 beta production toll-like receptor 3 signaling pathway positive regulation of cellular amide metabolic process response to isolation stress response to macrophage colony-stimulating factor NIK/NF-kappaB signaling vasodilation negative regulation of apoptotic process positive regulation of MAPK cascade negative regulation of blood vessel endothelial cell migration innate immune response response to ethanol negative regulation of fat cell differentiation positive regulation of protein catabolic process positive regulation of lipid metabolic process astrocyte activation regulation of inflammatory response negative regulation of neurogenesis regulation of developmental process modulation of synaptic transmission cognition regulation of secretion regulation of multicellular organismal process regulation of synaptic transmission, glutamatergic negative regulation of vascular wound healing regulation of biological quality positive regulation of ERK1 and ERK2 cascade cellular response to molecule of bacterial origin cellular response to retinoic acid cellular response to interferon-gamma cellular response to ionizing radiation positive regulation of mononuclear cell migration negative regulation of oxidative phosphorylation inflammatory response to wounding cellular response to toxic substance liver regeneration negative regulation of bile acid secretion antiviral innate immune response response to Gram-negative bacterium positive regulation of neuroinflammatory response positive regulation of interleukin-33 production response to L-glutamate positive regulation of neutrophil activation negative regulation of pri-miRNA transcription from RNA polymerase II promoter positive regulation of pri-miRNA transcription from RNA polymerase II promoter positive regulation of cellular protein localization cellular response to beta-amyloid positive regulation of leukocyte adhesion to vascular endothelial cell regulation of membrane lipid metabolic process response to 3,3',5-triiodo-L-thyronine positive regulation of hepatocyte proliferation regulation of endothelial cell apoptotic process positive regulation of DNA biosynthetic process negative regulation of apoptotic signaling pathway positive regulation of extrinsic apoptotic signaling pathway
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline