Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MRRGPRSLR GRDAPAPTPC VPAECFDLLV RHCVACGLLR TPRPKPAGAS SPAPRTALQP QESVGAGAGE AALPLPGLLF GAPALLGLAL VLALVLVGLV SWRRRQRRLR GASSAEAPDG DKDAPEPLDK VIILSPGISD ATAPAWPPPG EDPGTTPPGH SVPVPATELG STELVTTKTA GPEQQ (1-184 aa encoded by BC105123)
Tumor necrosis factor receptor superfamily member 13C (TNFRSF13C), also known as BAFFR, is a protein in humans is encoded by the TNFRSF13C gene. The BAFFR gene is mapped to chromosome 22q13. 1-q13. 31. It has got 184 amino acid transmembrane protein which is 56% identical to the mouse protein. B cell-activating factor (BAFF) enhances B-cell survival in vitro and is a regulator of the peripheral B-cell population. BAFF plays a crucial role in B cell development and can function through receptors other than BCMA.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: B cell-activating factor receptor; BAFF receptor; BAFF-R; BLyS receptor 3
Gene Aliases: BAFF-R; BAFFR; BROMIX; CD268; CVID4; prolixin
UniProt ID: (Human) Q96RJ3
Entrez Gene ID: (Human) 115650
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support