Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Predicted to react with Human, Mouse, and Rat samples.
TRAPPC1 plays a role in vesicular transport of proteins to the Golgi apparatus from the endoplasmic reticulum. The encoded protein is a component of the multisubunit transport protein particle (TRAPP) complex. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BET5 homolog; melanoma ubiquitous mutated 2; multiple myeloma protein 2; MUM-2; novel Sybindin-like family domain-containing protein; trafficking protein particle complex 1; unnamed protein product
Gene Aliases: BET5; D11Ertd172e; MUM2
UniProt ID: (Human) Q9Y5R8, (Mouse) Q5NCF2, (Rat) Q2KMM2
Entrez Gene ID: (Human) 58485, (Mouse) 245828, (Rat) 287427
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support